Search
Search

Rbpj| Gene Symbol: Rbpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106631.1 | NP_001100101.1 | 8 | 748 | ||
| XM_008770190.1 | XP_008768412.1 | 7 | 1419 | |||
| XM_008770191.1 | XP_008768413.1 | 7 | 660 | |||
| XM_008770192.1 | XP_008768414.1 | 7 | 2838 | |||
| XM_008770193.1 | XP_008768415.1 | 8 | 831 | |||