Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SP140 SIRNA Assays ›
          Gene Symbol:
          SP140
          Gene Name:
          SP140 nuclear body protein
          Gene Aliases:
          LYSP100, LYSP100-A, LYSP100-B
          Chromosome Location:
          Chr.2: 230202924 - 230316571 on Build GRCh38
          UniGene ID:
          Hs.632549
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22238

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SP140
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005176.2 NP_001005176.1 3 496
          NM_001278451.1 NP_001265380.1 3 496
          NM_001278452.1 NP_001265381.1 3 496
          NM_001278453.1 NP_001265382.1 3 496
          NM_007237.4 NP_009168.4 3 496
          XM_005246252.2 XP_005246309.1 3 460
          XM_005246253.2 XP_005246310.1 3 460
          XM_005246254.2 XP_005246311.1 3 460
          XM_005246255.2 XP_005246312.1 3 460
          XM_005246256.2 XP_005246313.1 3 460
          XM_006712223.3 XP_006712286.1 3 460
          XM_011510515.1 XP_011508817.1 3 460
          XM_011510516.1 XP_011508818.1 3 460
          XM_011510517.2 XP_011508819.1 5 1324
          XM_011510518.2 XP_011508820.1 4 596
          XM_011510519.1 XP_011508821.1 3 460
          XM_011510520.1 XP_011508822.1 3 460
          XM_017003239.1 XP_016858728.1 3 460
          XM_017003240.1 XP_016858729.1 3 460
          XM_017003241.1 XP_016858730.1 3 460
          XM_017003242.1 XP_016858731.1 3 460
          XM_017003243.1 XP_016858732.1 3 460
          XM_017003244.1 XP_016858733.1 4 596
          XM_017003245.1 XP_016858734.1 3 460
          XM_017003246.1 XP_016858735.1 3 460
          XM_017003247.1 XP_016858736.1 3 460
          XM_017003248.1 XP_016858737.1 3 460
          XM_017003249.1 XP_016858738.1 3 460
          XM_017003250.1 XP_016858739.1 3 460
          XM_017003251.1 XP_016858740.1 4 603
          XM_017003252.1 XP_016858741.1 3 460
          GenBank mRNA AK301267.1 - 3 496
          AK301317.1 - 3 497
          BC038091.1 - 3 467
          BC054890.1 - 3 466
          BC070160.1 - 3 462
          BC105743.1 - 3 479 40035747
          BC105744.1 - 3 481 40035746
          BC105960.1 - 3 481 40035752
          DC391925.1 - 3 496
          U36499.1 - 3 497
          U36500.1 - 3 497
          U63420.1 - 3 487

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          defense response
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0