Search
Search

Glyctk| Gene Symbol: Glyctk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109449.2 | NP_001102919.1 | 5 | 896 | ||
| XM_006243856.2 | XP_006243918.1 | 5 | 1171 | |||
| XM_006243857.2 | XP_006243919.1 | 5 | 954 | |||
| GenBank mRNA | BC105621.1 | - | 5 | 896 | 7454021 | |