Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AKT2 SIRNA Assays ›
          Gene Symbol:
          AKT2
          Gene Name:
          AKT serine/threonine kinase 2
          Gene Aliases:
          HIHGHH, PKBB, PKBBETA, PRKBB, RAC-BETA
          Chromosome Location:
          Chr.19: 40230317 - 40285531 on Build GRCh38
          UniGene ID:
          Hs.631535
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s223500

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AKT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243027.2 NP_001229956.1 5 692
          NM_001243028.2 NP_001229957.1 4 599
          NM_001626.5 NP_001617.1 5 729
          XM_011526614.1 XP_011524916.1 5 532
          XM_011526615.1 XP_011524917.1 5 609
          XM_011526616.1 XP_011524918.1 5 852
          XM_011526618.1 XP_011524920.1 5 694
          XM_011526619.1 XP_011524921.1 5 663
          XM_011526620.1 XP_011524922.1 5 598
          XM_011526622.2 XP_011524924.1 5 676
          XM_017026470.1 XP_016881959.1 6 811
          XM_017026471.1 XP_016881960.1 5 676
          GenBank mRNA AK055779.1 - 1 758
          AK294453.1 - 5 639
          AK304209.1 - 5 738
          AK308006.1 - 4 468
          AK308906.1 - 4 546
          AK314619.1 - 5 671
          BC032709.1 - 5 583
          BC040028.1 - 5 694
          BC120994.1 - 4 475 40116490
          BC120995.1 - 4 475 40116493
          KU159425.1 - 5 676
          M77198.1 - 5 633
          M95936.1 - 4 517

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          glycogen biosynthetic process
          glucose metabolic process
          regulation of translation
          cellular protein modification process
          apoptotic process
          response to osmotic stress
          signal transduction
          insulin receptor signaling pathway
          carbohydrate transport
          positive regulation of signal transduction
          negative regulation of plasma membrane long-chain fatty acid transport
          positive regulation of glucose metabolic process
          response to muscle activity
          peptidyl-serine phosphorylation
          regulation of cell migration
          positive regulation of cell migration
          positive regulation of vesicle fusion
          response to nutrient levels
          positive regulation of fatty acid beta-oxidation
          peripheral nervous system myelin maintenance
          cellular response to insulin stimulus
          negative regulation of RNA splicing
          positive regulation of peptidyl-serine phosphorylation
          intracellular signal transduction
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          protein kinase B signaling
          positive regulation of nitric oxide biosynthetic process
          fat cell differentiation
          positive regulation of glycogen biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of glucose import
          positive regulation of positive chemotaxis
          mammary gland epithelial cell differentiation
          intracellular protein transmembrane transport
          regulation of cell cycle arrest
          cellular response to organic cyclic compound
          protein localization to plasma membrane
          positive regulation of protein targeting to membrane
          activation of GTPase activity
          positive regulation of cell motility
          positive regulation of glucose import in response to insulin stimulus
          protein serine/threonine kinase activity
          protein kinase C binding
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0