Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLASP2 SIRNA Assays ›
Gene Symbol:
CLASP2
Gene Name:
cytoplasmic linker associated protein 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 33496245 - 33718356 on Build GRCh38
UniGene ID:
Hs.108614
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223572
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLASP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001207044.1
NP_001193973.1
29
3428
NM_015097.2
NP_055912.2
35
3850
XM_006713040.1
XP_006713103.1
37
4107
XM_006713041.1
XP_006713104.1
37
4104
XM_006713042.1
XP_006713105.1
36
4083
XM_006713048.1
XP_006713111.1
36
4044
XM_006713049.1
XP_006713112.1
35
4029
XM_006713050.1
XP_006713113.1
35
4020
XM_006713052.1
XP_006713115.1
35
3990
XM_006713053.1
XP_006713116.1
34
3969
XM_011533515.1
XP_011531817.1
35
4065
XM_017005947.1
XP_016861436.1
36
4080
XM_017005948.1
XP_016861437.1
35
4077
XM_017005949.1
XP_016861438.1
35
3908
XM_017005950.1
XP_016861439.1
36
4056
XM_017005951.1
XP_016861440.1
36
4053
XM_017005952.1
XP_016861441.1
35
4053
XM_017005953.1
XP_016861442.1
35
4050
XM_017005954.1
XP_016861443.1
36
4041
XM_017005955.1
XP_016861444.1
35
4032
XM_017005956.1
XP_016861445.1
34
4029
XM_017005957.1
XP_016861446.1
34
4017
XM_017005958.1
XP_016861447.1
34
3990
XM_017005959.1
XP_016861448.1
34
3987
XM_017005960.1
XP_016861449.1
33
3978
XM_017005961.1
XP_016861450.1
33
3821
XM_017005962.1
XP_016861451.1
34
3966
XM_017005963.1
XP_016861452.1
33
3966
XM_017005964.1
XP_016861453.1
33
3963
XM_017005965.1
XP_016861454.1
33
3951
XM_017005966.1
XP_016861455.1
32
3927
XM_017005967.1
XP_016861456.1
32
3770
XM_017005968.1
XP_016861457.1
30
3184
XM_017005969.1
XP_016861458.1
30
3194
XM_017005970.1
XP_016861459.1
30
3443
XM_017005971.1
XP_016861460.1
29
3170
XM_017005972.1
XP_016861461.1
30
3771
XM_017005973.1
XP_016861462.1
29
3416
XM_017005974.1
XP_016861463.1
28
3413
XM_017005975.1
XP_016861464.1
29
3133
XM_017005976.1
XP_016861465.1
27
3128
XM_017005977.1
XP_016861466.1
29
3392
XM_017005978.1
XP_016861467.1
28
3109
XM_017005979.1
XP_016861468.1
28
3119
XM_017005980.1
XP_016861469.1
29
3720
XM_017005981.1
XP_016861470.1
27
3106
XM_017005982.1
XP_016861471.1
27
3116
XM_017005983.1
XP_016861472.1
28
3368
XM_017005984.1
XP_016861473.1
28
3365
XM_017005985.1
XP_016861474.1
27
3693
XM_017005986.1
XP_016861475.1
26
3326
GenBank mRNA
AB014527.2
-
29
3067
AK297322.1
-
29
3103
AL137636.1
-
2
164
BC011530.1
-
15
1665
BC140778.1
-
35
3686
9021646
BX640725.1
-
11
1222
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
vesicle targeting
microtubule nucleation
negative regulation of microtubule depolymerization
Golgi organization
mitotic spindle organization
sister chromatid cohesion
establishment or maintenance of cell polarity
exit from mitosis
regulation of gastrulation
positive regulation of epithelial cell migration
regulation of epithelial to mesenchymal transition
microtubule organizing center organization
regulation of microtubule polymerization or depolymerization
regulation of microtubule-based process
microtubule anchoring
positive regulation of exocytosis
cell division
negative regulation of stress fiber assembly
negative regulation of focal adhesion assembly
establishment of protein localization to plasma membrane
positive regulation of extracellular matrix disassembly
negative regulation of wound healing, spreading of epidermal cells
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
dystroglycan binding
protein binding
microtubule binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-blthd:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline