Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARHGDIA SIRNA Assays ›
          Gene Symbol:
          ARHGDIA
          Gene Name:
          Rho GDP dissociation inhibitor alpha
          Gene Aliases:
          GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
          Chromosome Location:
          Chr.17: 81867719 - 81871406 on Build GRCh38
          UniGene ID:
          Hs.159161
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s224013

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARHGDIA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001185077.2 NP_001172006.1 6 834
          NM_001301240.1 NP_001288169.1 6 604
          NM_001301241.1 NP_001288170.1 6 834
          NM_001301243.1 NP_001288172.1 5 739
          NM_004309.5 NP_004300.1 6 604
          NR_125441.1 5 527
          XM_011523574.1 XP_011521876.1 5 964
          GenBank mRNA AF498926.1 - 5 468
          AK312347.1 - 6 540
          BC005851.2 - 6 560 2961556
          BC005875.2 - 6 560 2961556
          BC008701.3 - 5 495 2966760
          BC009759.2 - 6 525 3935956
          BC016031.1 - 5 476 3911525
          BC016185.1 - 6 520 4867857
          BC024258.1 - 6 520 3027721
          BC027730.2 - 6 523 4764249
          BC075827.1 - 6 520 6074977
          BC106044.1 - 6 563 6162284
          BF343803.1 - 63
          BI755343.1 - 6 526
          BQ674863.1 - 6 518
          BT006884.1 - 5 468
          CF132249.1 - 5 663
          CR456777.1 - 5 468
          D13989.1 - 5 488
          DA433004.1 - 5 446
          EU794615.1 - 5 495
          KU177916.1 - 5 468
          M97579.1 - 5 468
          X69550.1 - 6 521

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          negative regulation of cell adhesion
          regulation of Rho protein signal transduction
          negative regulation of apoptotic process
          positive regulation of GTPase activity
          negative regulation of axonogenesis
          positive regulation of axonogenesis
          regulation of small GTPase mediated signal transduction
          semaphorin-plexin signaling pathway
          regulation of actin cytoskeleton reorganization
          Rho GDP-dissociation inhibitor activity
          GTPase activator activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0