Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NTRK1 SIRNA Assays ›
          Gene Symbol:
          NTRK1
          Gene Name:
          neurotrophic receptor tyrosine kinase 1
          Gene Aliases:
          MTC, TRK, TRK1, TRKA, Trk-A, p140-TrkA
          Chromosome Location:
          Chr.1: 156815750 - 156881850 on Build GRCh38
          UniGene ID:
          Hs.406293
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s224140

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NTRK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007792.1 NP_001007793.1 15 1959
          NM_001012331.1 NP_001012331.1 14 2025
          NM_002529.3 NP_002520.2 15 2043
          GenBank mRNA AK126428.1 - 14 1897
          AK290759.1 - 14 2019
          AK312704.1 - 15 2028
          BC062580.1 - 15 1986
          BC136554.1 - 14 2069 9020544
          BC144239.1 - 14 2069 9052759
          CR936794.1 - 15 2442
          KJ534902.1 - 14 1969
          KJ534903.1 - 11 1423
          M23102.1 - 14 2089

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          positive regulation of protein phosphorylation
          protein phosphorylation
          microtubule-based movement
          axon guidance
          aging
          learning or memory
          circadian rhythm
          negative regulation of cell proliferation
          response to radiation
          programmed cell death involved in cell development
          positive regulation of neuron projection development
          response to activity
          peptidyl-tyrosine phosphorylation
          olfactory nerve development
          B cell differentiation
          response to nutrient levels
          nerve growth factor signaling pathway
          mechanoreceptor differentiation
          response to drug
          positive regulation of programmed cell death
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          response to ethanol
          positive regulation of Ras protein signal transduction
          protein autophosphorylation
          neurotrophin TRK receptor signaling pathway
          ephrin receptor signaling pathway
          phosphatidylinositol-mediated signaling
          sympathetic nervous system development
          response to axon injury
          detection of temperature stimulus involved in sensory perception of pain
          detection of mechanical stimulus involved in sensory perception of pain
          positive regulation of NF-kappaB transcription factor activity
          response to hydrostatic pressure
          response to electrical stimulus
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          Sertoli cell development
          axonogenesis involved in innervation
          behavioral response to formalin induced pain
          positive regulation of ERK1 and ERK2 cascade
          cellular response to nicotine
          cellular response to nerve growth factor stimulus
          transmembrane receptor protein tyrosine kinase activity
          GPI-linked ephrin receptor activity
          neurotrophin p75 receptor binding
          protein binding
          ATP binding
          nerve growth factor receptor activity
          kinase binding
          protein homodimerization activity
          neurotrophin binding
          nerve growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0