Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
Gene Name:
cAMP regulated phosphoprotein 21
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
UniGene ID:
Hs.475902
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s225449
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025068.1
NP_001020239.1
5
505
NM_001025069.1
NP_001020240.1
5
514
NM_001267616.1
NP_001254545.1
5
1661
NM_001267617.1
NP_001254546.1
5
471
NM_001267618.1
NP_001254547.1
4
602
NM_001267619.1
NP_001254548.1
4
436
NM_016300.4
NP_057384.2
5
488
NM_198399.1
NP_938409.1
4
496
XM_005264811.2
XP_005264868.1
5
517
XM_006712943.2
XP_006713006.1
5
517
XM_006712944.2
XP_006713007.1
5
517
XM_011533298.2
XP_011531600.1
5
517
XM_011533299.2
XP_011531601.1
5
1061
XM_011533300.2
XP_011531602.1
5
680
XM_011533301.2
XP_011531603.1
5
511
XM_011533302.2
XP_011531604.1
6
603
XM_011533303.2
XP_011531605.1
6
1153
XM_017005574.1
XP_016861063.1
5
607
XM_017005575.1
XP_016861064.1
6
12240
XM_017005576.1
XP_016861065.1
5
1061
XM_017005577.1
XP_016861066.1
5
511
XM_017005578.1
XP_016861067.1
5
487
XM_017005579.1
XP_016861068.1
5
1061
XM_017005580.1
XP_016861069.1
6
1153
XM_017005581.1
XP_016861070.1
6
12240
XM_017005582.1
XP_016861071.1
5
1061
XM_017005583.1
XP_016861072.1
5
511
XM_017005584.1
XP_016861073.1
5
1061
XM_017005585.1
XP_016861074.1
5
1061
XM_017005586.1
XP_016861075.1
5
517
XM_017005587.1
XP_016861076.1
5
680
XM_017005588.1
XP_016861077.1
6
1153
XM_017005589.1
XP_016861078.1
6
603
XM_017005590.1
XP_016861079.1
6
12240
XM_017005591.1
XP_016861080.1
5
1061
XM_017005592.1
XP_016861081.1
5
1061
XM_017005593.1
XP_016861082.1
5
607
XM_017005594.1
XP_016861083.1
5
511
XM_017005595.1
XP_016861084.1
6
1153
XM_017005596.1
XP_016861085.1
5
1061
XM_017005597.1
XP_016861086.1
5
1061
XM_017005598.1
XP_016861087.1
6
1153
XM_017005599.1
XP_016861088.1
5
1061
XM_017005600.1
XP_016861089.1
5
680
XM_017005601.1
XP_016861090.1
5
511
XM_017005602.1
XP_016861091.1
5
1061
XM_017005603.1
XP_016861092.1
5
680
XM_017005604.1
XP_016861093.1
5
511
XM_017005605.1
XP_016861094.1
6
1153
XM_017005606.1
XP_016861095.1
6
603
XM_017005607.1
XP_016861096.1
5
1061
XM_017005608.1
XP_016861097.1
5
1061
XM_017005609.1
XP_016861098.1
6
1153
XM_017005610.1
XP_016861099.1
5
1061
XM_017005611.1
XP_016861100.1
5
1061
XM_017005612.1
XP_016861101.1
5
1061
GenBank mRNA
AF112220.1
-
4
461
AK055064.1
-
5
514
AK289649.1
-
4
451
AK289868.1
-
5
508
AK294484.1
-
4
449
AV718064.1
-
5
505
AV718166.1
-
5
514
BC017805.1
-
4
496
BC036399.1
-
5
476
5263894
BC041385.1
-
5
1249
BC051828.1
-
5
488
6012704
BC066651.1
-
3
1015
BG535216.1
-
4
498
BX537647.1
-
1
618
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0