Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD6 SIRNA Assays ›
Gene Symbol:
ANKRD6
Gene Name:
ankyrin repeat domain 6
Gene Aliases:
-
Chromosome Location:
Chr.6: 89433178 - 89633834 on Build GRCh38
UniGene ID:
Hs.702213
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22579
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242809.1
NP_001229738.1
12
1438
NM_001242811.1
NP_001229740.1
12
1821
NM_001242813.1
NP_001229742.1
11
1333
NM_001242814.1
NP_001229743.1
9
1059
NM_014942.4
NP_055757.3
12
1438
XM_005248679.3
XP_005248736.1
11
1306
XM_005248680.3
XP_005248737.1
13
3610
XM_005248681.2
XP_005248738.1
12
1539
XM_005248682.3
XP_005248739.1
10
1228
XM_005248683.3
XP_005248740.1
9
1108
XM_005248684.3
XP_005248741.1
6
826
XM_011535596.2
XP_011533898.1
10
1207
XM_011535597.2
XP_011533899.1
10
1207
XM_011535598.2
XP_011533900.1
11
1308
XM_011535599.2
XP_011533901.1
11
1308
XM_011535600.2
XP_011533902.1
10
1230
XM_017010493.1
XP_016865982.1
10
1201
XM_017010494.1
XP_016865983.1
10
1207
XM_017010495.1
XP_016865984.1
11
1308
XM_017010496.1
XP_016865985.1
11
1472
XM_017010497.1
XP_016865986.1
10
1141
XM_017010498.1
XP_016865987.1
11
1302
XM_017010499.1
XP_016865988.1
9
1102
XM_017010500.1
XP_016865989.1
9
1108
XM_017010501.1
XP_016865990.1
10
1209
XM_017010502.1
XP_016865991.1
10
2890
XM_017010503.1
XP_016865992.1
10
2894
XM_017010504.1
XP_016865993.1
9
1036
XM_017010505.1
XP_016865994.1
10
1203
XM_017010506.1
XP_016865995.1
9
1042
XM_017010507.1
XP_016865996.1
9
1042
XM_017010508.1
XP_016865997.1
9
2786
XM_017010509.1
XP_016865998.1
8
937
XM_017010510.1
XP_016865999.1
8
992
XM_017010511.1
XP_016866000.1
8
2695
XM_017010512.1
XP_016866001.1
7
927
XM_017010513.1
XP_016866002.1
7
1059
XM_017010514.1
XP_016866003.1
6
828
XM_017010515.1
XP_016866004.1
12
1323
GenBank mRNA
AB023174.1
-
11
1160
AK074189.1
-
3
361
AK092235.1
-
3
1013
AK096867.1
-
9
921
AK295731.1
-
12
1738
AK307792.1
-
12
1407
AL117504.1
-
9
839
BC042173.1
-
12
1403
5454509
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
kinase inhibitor
kinase modulator
non-receptor serine/threonine protein kinase
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
positive regulation of JNK cascade
negative regulation of canonical Wnt signaling pathway
positive regulation of Wnt signaling pathway, planar cell polarity pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0