Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EEF2KMT SIRNA Assays ›
          Gene Symbol:
          EEF2KMT FAM86B1 FAM86B2 FAM86B3P
          Gene Name:
          eukaryotic elongation factor 2 lysine methyltransferase
          family with sequence similarity 86 member B1
          family with sequence similarity 86 member B2
          family with sequence similarity 86, member A pseudogene
          Gene Aliases:
          FAM86A, SB153, eEF2-KMT
          -
          -
          -
          Chromosome Location:
          Chr.16: 5084298 - 5097820 on Build GRCh38
          Chromosome Location:
          Chr.8: 12182104 - 12194118 on Build GRCh38
          Chromosome Location:
          Chr.8: 12424419 - 12436418 on Build GRCh38
          Chromosome Location:
          Chr.8: 8228570 - 8244865 on Build GRCh38
          UniGene ID:
          Hs.406461
          Hs.675799
          Hs.458413
          Hs.656318
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s225823

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EEF2KMT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289029.1 NP_001275958.1 4 451
          NM_201400.3 NP_958802.1 6 634
          NM_201598.3 NP_963892.1 5 532
          XM_005255157.4 XP_005255214.1 5 526
          XM_005255158.4 XP_005255215.1 7 680
          XM_011522404.2 XP_011520706.1 6 599
          GenBank mRNA AL522535.3 - 4 373
          AY037162.1 - 5 500
          BC010084.2 - 6 554 2822323
          BG024981.1 - 5 499
          BQ921410.1 - 6 617
          CA495371.1 - 5 453
          Gene Symbol: FAM86B1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083537.1 NP_001077006.1 5 496
          XM_006716256.3 XP_006716319.1 6 591
          XM_006716257.3 XP_006716320.1 5 519
          XM_006716260.3 XP_006716323.1 4 405
          XM_011543842.2 XP_011542144.1 4 528
          XM_011543843.2 XP_011542145.1 5 467
          XM_017013916.1 XP_016869405.1 5 495
          XM_017013917.1 XP_016869406.1 4 416
          XM_017013918.1 XP_016869407.1 4 388
          GenBank mRNA BE729618.1 - 5 438
          DA552021.1 - 1 29
          Gene Symbol: FAM86B2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001137610.1 NP_001131082.1 6 553
          XM_005273603.4 XP_005273660.1 5 526
          XM_005273604.4 XP_005273661.1 4 445
          XM_005273605.4 XP_005273662.1 4 346
          XM_006716387.3 XP_006716450.1 5 547
          XM_011544618.2 XP_011542920.1 6 598
          XM_011544619.2 XP_011542921.1 5 565
          XM_011544622.2 XP_011542924.1 5 467
          XM_017013740.1 XP_016869229.1 4 528
          XM_017013741.1 XP_016869230.1 3 465
          XM_017013742.1 XP_016869231.1 3 447
          Gene Symbol: FAM86B3P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024361.1 6 588
          NR_024362.1 6 685
          GenBank mRNA AK304567.1 - 6 657
          BC035792.1 - 6 563

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-lysine trimethylation
          methylation
          protein binding
          protein-lysine N-methyltransferase activity
          methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0