Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPIPB6 SIRNA Assays ›
          Gene Symbol:
          NPIPB6 NPIPB15 NPIPB8 NPIPB9 PDXDC2P
          Gene Name:
          nuclear pore complex interacting protein family member B6
          nuclear pore complex interacting protein family member B15
          nuclear pore complex interacting protein family member B8
          nuclear pore complex interacting protein family member B9
          pyridoxal dependent decarboxylase domain containing 2, pseudogene
          Gene Aliases:
          NPIPB
          A-761H5.4, NPIPL2
          -
          -
          NPIP, PDXDC2
          Chromosome Location:
          Chr.16: 28342517 - 28364971 on Build GRCh38
          Chromosome Location:
          Chr.16: 74374986 - 74392115 on Build GRCh38
          Chromosome Location:
          Chr.16: 28638065 - 28658693 on Build GRCh38
          Chromosome Location:
          Chr.16: 28735500 - 28772911 on Build GRCh38
          Chromosome Location:
          Chr.16: 69976297 - 70065948 on Build GRCh38
          UniGene ID:
          Hs.726467
          -
          -
          Hs.710214
          Hs.513695
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s226611

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPIPB6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282524.1 NP_001269453.1 7 1187
          XM_005255741.3 XP_005255798.1 8 1999
          XM_005255743.3 XP_005255800.1 4 2578
          XM_011545929.2 XP_011544231.1 9 1819
          XM_011545931.2 XP_011544233.1 8 1452
          XM_017023628.1 XP_016879117.1 8 1202
          XM_017023629.1 XP_016879118.1 8, 9 1703
          XM_017023630.1 XP_016879119.1 9 1472
          XM_017023631.1 XP_016879120.1 7 1816
          XM_017023632.1 XP_016879121.1 8, 9 1160
          GenBank mRNA BC144439.1 - 7 1148
          BC157836.1 - 8 1900 9057055
          Gene Symbol: NPIPB15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306094.1 NP_001293023.1 7 1165
          XM_011523496.2 XP_011521798.1 8 1221
          XM_011523498.2 XP_011521800.1 8 1255
          XM_011523501.2 XP_011521803.1 4 1047
          XM_017023248.1 XP_016878737.1 8 1658
          XM_017023249.1 XP_016878738.1 7 1475
          Gene Symbol: NPIPB8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310136.1 NP_001297065.1 7 1170
          XM_017023622.1 XP_016879111.1 8 1441
          XM_017023623.1 XP_016879112.1 9 1378
          XM_017023624.1 XP_016879113.1 8 1294
          XM_017023625.1 XP_016879114.1 8 1274
          XM_017023626.1 XP_016879115.1 9 1913
          Gene Symbol: NPIPB9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001287250.1 NP_001274179.1 7 1208
          NM_001287251.1 NP_001274180.1 8 1294
          XM_003118719.7 XP_003118767.2 8 1769
          XM_011545690.2 XP_011543992.1 10 1930
          XM_011545693.2 XP_011543995.1 9 1857
          XM_017022814.1 XP_016878303.1 9 2005
          XM_017022815.1 XP_016878304.1 9 1525
          XM_017022816.1 XP_016878305.1 10 1942
          XM_017022817.1 XP_016878306.1 8 1917
          XM_017022818.1 XP_016878307.1 8 3242
          XM_017022819.1 XP_016878308.1 9 1932
          XM_017022820.1 XP_016878309.1 9 1649
          XM_017022821.1 XP_016878310.1 8 2515
          XM_017022822.1 XP_016878311.1 9 3518
          XM_017022823.1 XP_016878312.1 7 3154
          XM_017022824.1 XP_016878313.1 9 1784
          XM_017022825.1 XP_016878314.1 9 1502
          XM_017022826.1 XP_016878315.1 8 1466
          XM_017022827.1 XP_016878316.1 4 1213
          Gene Symbol: PDXDC2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BX647358.1 - 2 5813

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0