Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MED15 SIRNA Assays ›
Gene Symbol:
MED15
Gene Name:
mediator complex subunit 15
Gene Aliases:
ARC105, CAG7A, CTG7A, PCQAP, TIG-1, TIG1, TNRC7
Chromosome Location:
Chr.22: 20507542 - 20587632 on Build GRCh38
UniGene ID:
Hs.517421
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s226742
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MED15
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003891.2
NP_001003891.1
4, 5
376
NM_001293234.1
NP_001280163.1
4, 5
376
NM_001293235.1
NP_001280164.1
4, 5
365
NM_001293237.1
NP_001280166.1
4, 5
568
NM_015889.4
NP_056973.2
4, 5
376
XM_006724264.3
XP_006724327.1
4, 5
347
XM_011530214.2
XP_011528516.1
5, 6
420
XM_011530215.2
XP_011528517.1
3, 4
242
XM_011530218.2
XP_011528520.1
4, 5
376
XM_011530219.1
XP_011528521.1
4, 5
376
XM_011530220.2
XP_011528522.1
4, 5
376
XM_017028817.1
XP_016884306.1
3, 4
318
XM_017028818.1
XP_016884307.1
5, 6
518
XM_017028819.1
XP_016884308.1
4, 5
284
XM_017028820.1
XP_016884309.1
4, 5
347
XM_017028821.1
XP_016884310.1
4, 5
376
XM_017028822.1
XP_016884311.1
4, 5
376
XM_017028823.1
XP_016884312.1
3, 4
171
XM_017028824.1
XP_016884313.1
4, 5
347
GenBank mRNA
AF056191.1
-
4, 5
325
AF328769.1
-
4, 5
239
AK074268.1
-
4, 5
338
AK090465.1
-
4, 5
308
AK294540.1
-
4, 5
384
AK294734.1
-
4, 5
564
AK315540.1
-
4, 5
336
BC003078.1
-
4, 5
297
BC013985.2
-
4, 5
279
3506327
CB156495.1
-
4, 5
568
CB988805.1
-
4, 5
372
DA182752.1
-
4, 5
379
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
RNA polymerase II transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0