Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TSPY3 SIRNA Assays ›
          Gene Symbol:
          TSPY3 TSPY1 TSPY10 TSPY2 TSPY4 TSPY8
          Gene Name:
          testis specific protein, Y-linked 3
          testis specific protein, Y-linked 1
          testis specific protein, Y-linked 10
          testis specific protein, Y-linked 2
          testis specific protein, Y-linked 4
          testis specific protein, Y-linked 8
          Gene Aliases:
          CT78
          CT78, DYS14, TSPY, pJA923
          -
          TSPYQ1
          TSPY10, TSPY8
          -
          Chromosome Location:
          Chr.Y: 9398421 - 9401224 on Build GRCh38
          Chromosome Location:
          Chr.Y: 9466955 - 9469756 on Build GRCh38
          Chromosome Location:
          Chr.Y: 9527880 - 9530676 on Build GRCh38
          Chromosome Location:
          Chr.Y: 6246223 - 6249020 on Build GRCh38
          Chromosome Location:
          Chr.Y: 9337489 - 9340285 on Build GRCh38
          Chromosome Location:
          Chr.Y: 9357829 - 9360600 on Build GRCh38
          UniGene ID:
          Hs.556121
          Hs.556121
          Hs.556121
          Hs.646249
          Hs.556121
          Hs.740264
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID s227476

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TSPY3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077697.2 NP_001071165.2 6 998
          XM_005272742.4 XP_005272799.1 6 984
          XM_011544004.2 XP_011542306.1 6 966
          Gene Symbol: TSPY1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001197242.2 NP_001184171.1 6 1009
          NM_001320964.1 NP_001307893.1 7 737
          NM_003308.4 NP_003299.2 6 998
          XM_017030064.1 XP_016885553.1 6 966
          GenBank mRNA BC121113.1 - 7 741 40120433
          M98525.1 - 6 913
          U58096.1 - 6 997
          Gene Symbol: TSPY10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282469.2 NP_001269398.1 6 998
          NM_001320962.1 NP_001307891.1 6 1009
          XM_017030025.1 XP_016885514.1 6 966
          GenBank mRNA AY130858.1 - 6 963
          BC121114.1 - 6 1002 40120435
          DB545400.1 - 4 332
          Gene Symbol: TSPY2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022573.2 NP_072095.2 6 998
          XM_005262569.4 XP_005262626.1 6 969
          XM_017030062.1 XP_016885551.1 6 951
          GenBank mRNA AK307425.1 - 6 903
          Gene Symbol: TSPY4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164471.1 NP_001157943.1 6 970
          XM_005272743.4 XP_005272800.1 6 1002
          XM_017030065.1 XP_016885554.1 6 984
          GenBank mRNA AK093413.1 - 4 1744
          BX281192.1 - 5 464
          Gene Symbol: TSPY8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243721.1 NP_001230650.1 6 952
          XM_005272744.4 XP_005272801.1 6 977
          XM_011544005.2 XP_011542307.1 6 959

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          spermatogenesis
          gonadal mesoderm development
          cell differentiation
          sex differentiation
          cell proliferation
          molecular_function
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pzqlg:80/100.66.77.245:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0