Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNA1C SIRNA Assays ›
          Gene Symbol:
          CACNA1C
          Gene Name:
          calcium voltage-gated channel subunit alpha1 C
          Gene Aliases:
          CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
          Chromosome Location:
          Chr.12: 1969677 - 2697949 on Build GRCh38
          UniGene ID:
          Hs.118262
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2285

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CACNA1C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000719.6 NP_000710.5 15, 16 2540
          NM_001129827.1 NP_001123299.1 15, 16 2540
          NM_001129829.1 NP_001123301.1 15, 16 2540
          NM_001129830.2 NP_001123302.2 15, 16 2505
          NM_001129831.1 NP_001123303.1 15, 16 2540
          NM_001129832.1 NP_001123304.1 15, 16 2540
          NM_001129833.1 NP_001123305.1 15, 16 2540
          NM_001129834.1 NP_001123306.1 15, 16 2540
          NM_001129835.1 NP_001123307.1 15, 16 2540
          NM_001129836.1 NP_001123308.1 15, 16 2540
          NM_001129837.1 NP_001123309.1 15, 16 2540
          NM_001129838.1 NP_001123310.1 15, 16 2540
          NM_001129839.1 NP_001123311.1 15, 16 2540
          NM_001129840.1 NP_001123312.1 15, 16 2540
          NM_001129841.1 NP_001123313.1 15, 16 2540
          NM_001129842.1 NP_001123314.1 15, 16 2540
          NM_001129843.1 NP_001123315.1 15, 16 2540
          NM_001129844.1 NP_001123316.1 15, 16 2531
          NM_001129846.1 NP_001123318.1 15, 16 2540
          NM_001167623.1 NP_001161095.1 15, 16 2540
          NM_001167624.2 NP_001161096.2 15, 16 2505
          NM_001167625.1 NP_001161097.1 15, 16 2540
          NM_199460.3 NP_955630.3 15, 16 2505
          XM_006719017.2 XP_006719080.1 15, 16 2594
          XM_011521020.2 XP_011519322.1 16, 17 2669
          XM_011521023.2 XP_011519325.1 16, 17 2854
          XM_017019926.1 XP_016875415.1 18, 19 3976
          XM_017019927.1 XP_016875416.1 18, 19 3976
          XM_017019928.1 XP_016875417.1 18, 19 3976
          XM_017019929.1 XP_016875418.1 18, 19 3976
          XM_017019930.1 XP_016875419.1 18, 19 3976
          XM_017019931.1 XP_016875420.1 18, 19 3976
          XM_017019932.1 XP_016875421.1 16, 17 3781
          XM_017019933.1 XP_016875422.1 16, 17 3781
          XM_017019934.1 XP_016875423.1 17, 18 3856
          XM_017019935.1 XP_016875424.1 17, 18 3856
          XM_017019936.1 XP_016875425.1 16, 17 3781
          XM_017019937.1 XP_016875426.1 16, 17 3781
          XM_017019938.1 XP_016875427.1 16, 17 3781
          XM_017019939.1 XP_016875428.1 16, 17 3781
          XM_017019940.1 XP_016875429.1 16, 17 3781
          XM_017019941.1 XP_016875430.1 16, 17 3781
          XM_017019942.1 XP_016875431.1 16, 17 3781
          XM_017019943.1 XP_016875432.1 16, 17 3781
          XM_017019944.1 XP_016875433.1 16, 17 3781
          XM_017019945.1 XP_016875434.1 16, 17 3781
          XM_017019946.1 XP_016875435.1 16, 17 3781
          XM_017019947.1 XP_016875436.1 16, 17 3781
          XM_017019948.1 XP_016875437.1 16, 17 3781
          XM_017019949.1 XP_016875438.1 16, 17 3781
          XM_017019950.1 XP_016875439.1 16, 17 3781
          XM_017019951.1 XP_016875440.1 16, 17 3772
          XM_017019952.1 XP_016875441.1 16, 17 3781
          XM_017019953.1 XP_016875442.1 15, 16 2594
          XM_017019954.1 XP_016875443.1 15, 16 2594
          XM_017019955.1 XP_016875444.1 17, 18 3856
          GenBank mRNA AB209016.1 - 13, 14 1975
          AF465484.1 - 15, 16 2492
          AJ224873.1 - 15, 16 2227
          AY830711.1 - 15, 16 2227
          AY830712.1 - 15, 16 2227
          AY830713.1 - 15, 16 2227
          BC146846.1 - 15, 16 2346 9056774
          L29529.1 - 14, 15 2159
          L29534.1 - 14, 15 2153
          L29536.1 - 14, 15 2153
          Z34809.1 - 15, 16 2227
          Z34810.1 - 15, 16 2227
          Z34811.1 - 15, 16 2227
          Z34812.1 - 15, 16 2227
          Z34813.1 - 15, 16 2227
          Z34814.1 - 15, 16 2227
          Z34815.1 - 15, 16 2227
          Z34816.1 - 15, 16 2227
          Z34817.1 - 15, 16 2227
          Z34818.1 - 15, 16 2227
          Z34819.1 - 15, 16 2227
          Z34820.1 - 15, 16 2227
          Z34821.1 - 15, 16 2227
          Z34822.1 - 15, 16 2227
          Z74996.1 - 15, 16 2227

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system development
          positive regulation of cytosolic calcium ion concentration
          heart development
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          embryonic forelimb morphogenesis
          calcium-mediated signaling using extracellular calcium source
          camera-type eye development
          regulation of insulin secretion
          calcium ion transport into cytosol
          cardiac conduction
          calcium ion transmembrane transport via high voltage-gated calcium channel
          calcium ion transmembrane transport
          cardiac muscle cell action potential involved in contraction
          membrane depolarization during cardiac muscle cell action potential
          membrane depolarization during AV node cell action potential
          cell communication by electrical coupling involved in cardiac conduction
          regulation of heart rate by cardiac conduction
          regulation of ventricular cardiac muscle cell action potential
          membrane depolarization during atrial cardiac muscle cell action potential
          voltage-gated calcium channel activity
          protein binding
          calmodulin binding
          high voltage-gated calcium channel activity
          metal ion binding
          alpha-actinin binding
          voltage-gated calcium channel activity involved in cardiac muscle cell action potential
          voltage-gated calcium channel activity involved in AV node cell action potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0