Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RUNX1 SIRNA Assays ›
          Gene Symbol:
          RUNX1
          Gene Name:
          runt related transcription factor 1
          Gene Aliases:
          AML1, AML1-EVI-1, AMLCR1, CBF2alpha, CBFA2, EVI-1, PEBP2aB, PEBP2alpha
          Chromosome Location:
          Chr.21: 34787801 - 35049334 on Build GRCh38
          UniGene ID:
          Hs.149261
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s229352

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RUNX1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001890.2 NP_001001890.1 2 1960
          NM_001122607.1 NP_001116079.1 2 1960
          NM_001754.4 NP_001745.2 5 653
          XM_005261068.3 XP_005261125.1 3 916
          XM_005261069.4 XP_005261126.1 5 689
          XM_011529766.2 XP_011528068.1 5 764
          XM_011529767.2 XP_011528069.1 4 650
          XM_011529768.2 XP_011528070.1 4 650
          XM_011529770.2 XP_011528072.1 5 689
          XM_017028487.1 XP_016883976.1 3 458
          GenBank mRNA AK226159.1 - 4 650
          AK310587.1 - 5 642
          BC110828.1 - 4 548
          BC136380.1 - 5 610 9020367
          BC136381.1 - 5 609 9020368
          BC144053.1 - 5 609 9052573
          D10570.1 - 2 1150
          D43967.1 - 2 1960
          D43968.1 - 2 1960
          D43969.1 - 4 907
          D89788.1 - 3 350
          D89789.1 - 3 350
          D89790.1 - 3 350
          L34598.1 - 2 1060
          S60998.1 - 2 390
          U19601.1 - 4 463
          X79549.1 - 2 1950

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          Runt transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          chondrocyte differentiation
          transcription from RNA polymerase II promoter
          hemopoiesis
          positive regulation of granulocyte differentiation
          positive regulation of interleukin-2 production
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          peripheral nervous system neuron development
          hematopoietic stem cell proliferation
          regulatory region DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          core promoter binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0