Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIP5K1A SIRNA Assays ›
          Gene Symbol:
          PIP5K1A PIPSL
          Gene Name:
          phosphatidylinositol-4-phosphate 5-kinase type 1 alpha
          PIP5K1A and PSMD4-like, pseudogene
          Gene Aliases:
          -
          PIP5K1L1, PIP5K1P3, PSMD4P2, bA429H9.1
          Chromosome Location:
          Chr.1: 151198003 - 151249536 on Build GRCh38
          Chromosome Location:
          Chr.10: 93958140 - 93961915 on Build GRCh38
          UniGene ID:
          Hs.655131
          Hs.149255
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s229449

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PIP5K1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135636.1 NP_001129108.1 7 1170
          NM_001135637.1 NP_001129109.1 7 1167
          NM_001135638.1 NP_001129110.1 8 1206
          NM_003557.2 NP_003548.1 7 1167
          XM_005245525.4 XP_005245582.1 9 1447
          XM_005245527.3 XP_005245584.1 8 1196
          XM_005245528.4 XP_005245585.1 8 1411
          XM_005245530.4 XP_005245587.1 8 769
          XM_005245531.3 XP_005245588.1 7 1163
          XM_005245532.3 XP_005245589.1 7 1096
          XM_006711563.3 XP_006711626.1 9 1453
          XM_006711564.3 XP_006711627.1 8 1165
          XM_006711567.3 XP_006711630.1 9 753
          XM_006711568.3 XP_006711631.2 8 784
          XM_011510043.2 XP_011508345.1 7 748
          XM_011510045.2 XP_011508347.1 8 766
          XM_011510046.2 XP_011508348.1 7 1241
          XM_011510048.2 XP_011508350.1 9 763
          XM_011510049.2 XP_011508351.1 9 750
          XM_017002439.1 XP_016857928.1 7 1163
          XM_017002440.1 XP_016857929.1 7 1160
          XM_017002441.1 XP_016857930.1 9 769
          GenBank mRNA AK291015.1 - 7 1110
          AK295691.1 - 7 1163
          BC007833.2 - 7 1159 4098514
          U78575.1 - 7 1115
          U78576.1 - 8 1175
          U78577.1 - 7 1116
          Gene Symbol: PIPSL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002319.2 1 1114

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycerophospholipid metabolic process
          phosphatidylinositol biosynthetic process
          phagocytosis
          signal transduction
          phospholipid biosynthetic process
          fibroblast migration
          regulation of phosphatidylinositol 3-kinase signaling
          cell migration
          keratinocyte differentiation
          actin cytoskeleton reorganization
          phosphatidylinositol phosphorylation
          focal adhesion assembly
          cell chemotaxis
          protein targeting to plasma membrane
          activation of GTPase activity
          ruffle assembly
          1-phosphatidylinositol-3-phosphate 5-kinase activity
          protein binding
          ATP binding
          1-phosphatidylinositol-4-phosphate 5-kinase activity
          kinase binding
          1-phosphatidylinositol-5-kinase activity
          1-phosphatidylinositol-3-phosphate 4-kinase activity
          phosphatidylinositol-3,4-bisphosphate 5-kinase activity
          phosphatidylinositol phosphate kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-wl76t:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0