Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAHD2A SIRNA Assays ›
          Gene Symbol:
          FAHD2A FAHD2B FAHD2CP
          Gene Name:
          fumarylacetoacetate hydrolase domain containing 2A
          fumarylacetoacetate hydrolase domain containing 2B
          fumarylacetoacetate hydrolase domain containing 2C, pseudogene
          Gene Aliases:
          CGI-105
          -
          -
          Chromosome Location:
          Chr.2: 95402691 - 95416702 on Build GRCh38
          Chromosome Location:
          Chr.2: 97080738 - 97094882 on Build GRCh38
          Chromosome Location:
          Chr.2: 96010551 - 96023136 on Build GRCh38
          UniGene ID:
          Hs.531247
          Hs.567723
          Hs.720108
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s229761

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAHD2A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016044.2 NP_057128.2 6 933
          XM_011511282.2 XP_011509584.1 6 1682
          XM_011511283.2 XP_011509585.1 7 1059
          XM_011511284.2 XP_011509586.1 7 1188
          XM_011511285.2 XP_011509587.1 7 1056
          XM_011511286.2 XP_011509588.1 6 942
          XM_011511287.2 XP_011509589.1 6 1053
          XM_011511288.2 XP_011509590.1 6 1682
          XM_011511289.2 XP_011509591.1 6 1682
          XM_011511290.2 XP_011509592.1 4 1459
          XM_017004236.1 XP_016859725.1 7 1059
          XM_017004237.1 XP_016859726.1 7 1188
          XM_017004238.1 XP_016859727.1 7 1056
          XM_017004239.1 XP_016859728.1 6 942
          XM_017004240.1 XP_016859729.1 6 1053
          XM_017004241.1 XP_016859730.1 6 1682
          XM_017004242.1 XP_016859731.1 6 942
          XM_017004243.1 XP_016859732.1 5 1519
          XM_017004244.1 XP_016859733.1 5 779
          XM_017004245.1 XP_016859734.1 5 1519
          XM_017004246.1 XP_016859735.1 5 1519
          XM_017004247.1 XP_016859736.1 5 779
          XM_017004248.1 XP_016859737.1 5 890
          XM_017004249.1 XP_016859738.1 4 710
          XM_017004250.1 XP_016859739.1 4 830
          GenBank mRNA AF151863.1 - 6 931
          BC009403.2 - 6 907 4309334
          BC110911.1 - 6 905 6064857
          Gene Symbol: FAHD2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320848.1 NP_001307777.1 7 1068
          NM_199336.2 NP_955368.1 6 942
          XM_011510745.2 XP_011509047.1 7 1058
          XM_011510746.2 XP_011509048.1 7 1177
          XM_011510747.2 XP_011509049.1 6 942
          XM_011510748.2 XP_011509050.1 7 1058
          XM_011510750.2 XP_011509052.1 5 837
          XM_017003470.1 XP_016858959.1 7 1055
          XM_017003471.1 XP_016858960.1 7 1058
          XM_017003472.1 XP_016858961.1 6 991
          XM_017003473.1 XP_016858962.1 4 719
          GenBank mRNA AL833869.1 - 5 742
          BC064511.1 - 6 905 6644987
          Gene Symbol: FAHD2CP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003698.1 2 251
          GenBank mRNA BI916267.1 - 2 218
          BM790702.1 - 2 251
          CN274361.1 - 2 184

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          hydrolase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0