Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CFLAR SIRNA Assays ›
          Gene Symbol:
          CFLAR
          Gene Name:
          CASP8 and FADD like apoptosis regulator
          Gene Aliases:
          CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS
          Chromosome Location:
          Chr.2: 201116015 - 201172688 on Build GRCh38
          UniGene ID:
          Hs.390736
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s229912

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CFLAR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127183.2 NP_001120655.1 4 958
          NM_001127184.2 NP_001120656.1 4 930
          NM_001202516.1 NP_001189445.1 4 930
          NM_001202517.1 NP_001189446.1 3 512
          NM_001202518.1 NP_001189447.1 2 248
          NM_001202519.1 NP_001189448.1 3 512
          NM_001308042.1 NP_001294971.1 3 602
          NM_001308043.1 NP_001294972.1 4 980
          NM_003879.5 NP_003870.4 4 930
          XM_017005189.1 XP_016860678.1 4 921
          XM_017005190.1 XP_016860679.1 4 794
          XM_017005191.1 XP_016860680.1 4 873
          XM_017005192.1 XP_016860681.1 4 921
          XM_017005193.1 XP_016860682.1 4 2938
          XM_017005194.1 XP_016860683.1 4 794
          XM_017005195.1 XP_016860684.1 5 971
          XM_017005196.1 XP_016860685.1 3 651
          XM_017005197.1 XP_016860686.1 4 2938
          XM_017005198.1 XP_016860687.1 4 794
          GenBank mRNA AB209600.1 - 4 677
          AF005774.1 - 4 900
          AF005775.1 - 3 465
          AF009616.1 - 4 877
          AF009617.1 - 3 465
          AF009618.1 - 3 465
          AF009619.1 - 3 465
          AF010127.1 - 4 875
          AF015450.1 - 4 969
          AF015451.1 - 3 465
          AF015452.1 - 3 465
          AF041458.1 - 3 480
          AF041460.1 - 3 465
          AF041461.1 - 3 420
          AF041462.1 - 2 177
          AK289913.1 - 4 754
          AK296036.1 - 3 512
          AK297387.1 - 5 827
          AK304585.1 - 2 229
          AK308767.1 - 4 840
          AK315208.1 - 4 701
          AK315924.1 - 3 316
          BC001602.1 - 4 724 3542993
          BT006751.1 - 3 465
          CD699251.1 - 2 281
          U85059.1 - 3 465
          U97074.1 - 4 847
          U97075.1 - 3 758
          Y14039.1 - 4 946
          Y14040.1 - 4 946

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          apoptotic process
          skeletal muscle tissue development
          skeletal muscle atrophy
          regulation of skeletal muscle satellite cell proliferation
          skeletal myofibril assembly
          viral process
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          skeletal muscle tissue regeneration
          positive regulation of NF-kappaB transcription factor activity
          regulation of necroptotic process
          execution phase of apoptosis
          negative regulation of myoblast fusion
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway
          protease binding
          death receptor binding
          protein binding
          enzyme activator activity
          protein complex binding
          cysteine-type endopeptidase activity involved in execution phase of apoptosis

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0