Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNB2 SIRNA Assays ›
Gene Symbol:
CACNB2
Gene Name:
calcium voltage-gated channel auxiliary subunit beta 2
Gene Aliases:
CACNLB2, CAVB2, MYSB
Chromosome Location:
Chr.10: 18140560 - 18545881 on Build GRCh38
UniGene ID:
Hs.59093
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2305
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000724.3
NP_000715.2
3
805
NM_001167945.1
NP_001161417.1
4
638
NM_201570.2
NP_963864.1
3
659
NM_201571.3
NP_963865.2
4
638
NM_201572.3
NP_963866.2
4
638
NM_201590.2
NP_963884.2
3
435
NM_201593.2
NP_963887.2
4
415
NM_201596.2
NP_963890.2
4
415
NM_201597.2
NP_963891.1
4
415
XM_005252588.3
XP_005252645.1
3
663
XM_006717502.3
XP_006717565.1
3
468
XM_011519659.2
XP_011517961.1
3
435
XM_011519660.2
XP_011517962.1
3
1020
XM_017016625.1
XP_016872114.1
3
575
GenBank mRNA
AB208917.1
-
3
691
AF137377.1
-
3
196
AF285239.2
-
3
193
AF423189.1
-
3
190
AF423190.1
-
4
271
AF423191.1
-
4
355
AF423192.1
-
3
211
AF465485.1
-
3
199
AK293837.1
-
4
570
AK293937.1
-
3
220
AK295113.1
-
3
556
AK296471.1
-
4
322
AK299873.1
-
3
561
AK307526.1
-
4
638
AK307800.1
-
3
381
AK310943.1
-
3
435
AL119895.1
-
4
303
AL600557.1
-
3
360
AY393858.1
-
4
271
AY393859.1
-
3
193
AY393860.1
-
4
355
AY393861.1
-
4
271
AY393862.1
-
3
193
AY393863.1
-
3
193
AY675091.1
-
4
355
BC136409.1
-
4
378
9020397
DB235418.1
-
3
435
S60415.1
-
4
415
U95019.1
-
3
691
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
neuromuscular junction development
visual perception
positive regulation of calcium ion transport
cardiac conduction
calcium ion import
calcium ion transmembrane transport
membrane depolarization during AV node cell action potential
regulation of heart rate by cardiac conduction
establishment of protein localization to plasma membrane
membrane depolarization during atrial cardiac muscle cell action potential
regulation of voltage-gated calcium channel activity
positive regulation of high voltage-gated calcium channel activity
positive regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
voltage-gated calcium channel activity
calcium channel regulator activity
calcium channel activity
protein binding
high voltage-gated calcium channel activity
protein kinase binding
protein domain specific binding
actin filament binding
phosphoprotein binding
voltage-gated calcium channel activity involved in cardiac muscle cell action potential
voltage-gated calcium channel activity involved in AV node cell action potential
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0