Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNB2 SIRNA Assays ›
          Gene Symbol:
          CACNB2
          Gene Name:
          calcium voltage-gated channel auxiliary subunit beta 2
          Gene Aliases:
          CACNLB2, CAVB2, MYSB
          Chromosome Location:
          Chr.10: 18140560 - 18545881 on Build GRCh38
          UniGene ID:
          Hs.59093
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2306

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CACNB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000724.3 NP_000715.2 10, 11 1664
          NM_001167945.1 NP_001161417.1 11, 12 1383
          NM_201570.2 NP_963864.1 10, 11 1518
          NM_201571.3 NP_963865.2 11, 12 1497
          NM_201572.3 NP_963866.2 11, 12 1425
          NM_201590.2 NP_963884.2 10, 11 1294
          NM_201593.2 NP_963887.2 11, 12 1160
          NM_201596.2 NP_963890.2 11, 12 1274
          NM_201597.2 NP_963891.1 11, 12 1202
          XM_005252588.3 XP_005252645.1 10, 11 1408
          XM_005252591.3 XP_005252648.1 5, 6 469
          XM_006717502.3 XP_006717565.1 10, 11 1327
          XM_011519659.2 XP_011517961.1 10, 11 1222
          XM_011519660.2 XP_011517962.1 10, 11 1765
          XM_017016625.1 XP_016872114.1 11, 12 1382
          GenBank mRNA AB208917.1 - 10, 11 1550
          AF285239.2 - 10, 11 1052
          AF423189.1 - 10, 11 1049
          AF423190.1 - 11, 12 1130
          AF423191.1 - 11, 12 1214
          AF423192.1 - 10, 11 1070
          AF465485.1 - 10, 11 944
          AK293937.1 - 10, 11 965
          AK296471.1 - 11, 12 1181
          AK299873.1 - 10, 11 1420
          AK307526.1 - 11, 12 1383
          AY393858.1 - 11, 12 1130
          AY393859.1 - 10, 11 1052
          AY393860.1 - 11, 12 1142
          AY393861.1 - 11, 12 1058
          AY393862.1 - 10, 11 980
          BC136409.1 - 11, 12 1237 9020397
          S60415.1 - 11, 12 1274
          U95019.1 - 10, 11 1550

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053851, XM_006254304, XM_006254303, XM_006254300, XM_006254301, and others.

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          neuromuscular junction development
          visual perception
          positive regulation of calcium ion transport
          cardiac conduction
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during AV node cell action potential
          regulation of heart rate by cardiac conduction
          establishment of protein localization to plasma membrane
          membrane depolarization during atrial cardiac muscle cell action potential
          regulation of voltage-gated calcium channel activity
          positive regulation of high voltage-gated calcium channel activity
          positive regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
          voltage-gated calcium channel activity
          calcium channel regulator activity
          calcium channel activity
          protein binding
          high voltage-gated calcium channel activity
          protein kinase binding
          protein domain specific binding
          actin filament binding
          phosphoprotein binding
          voltage-gated calcium channel activity involved in cardiac muscle cell action potential
          voltage-gated calcium channel activity involved in AV node cell action potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0