Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WASH3P SIRNA Assays ›
          Gene Symbol:
          WASH3P LOC100288778 WASH1 WASH2P WASH7P
          Gene Name:
          WAS protein family homolog 3 pseudogene
          WAS protein family homolog 1 pseudogene
          WAS protein family homolog 1
          WAS protein family homolog 2 pseudogene
          WAS protein family homolog 7 pseudogene
          Gene Aliases:
          FAM39DP
          -
          FAM39E, WASH
          FAM39B
          FAM39F, WASH5P
          Chromosome Location:
          Chr.15: 101960813 - 101976605 on Build GRCh38
          Chromosome Location:
          Chr.12: 14477 - 32015 on Build GRCh38
          Chromosome Location:
          Chr.9: 14475 - 30487 on Build GRCh38
          Chromosome Location:
          Chr.2: 113583653 - 113599037 on Build GRCh38
          Chromosome Location:
          Chr.1: 14362 - 29370 on Build GRCh38
          UniGene ID:
          Hs.459573
          Hs.459573
          Hs.446466
          Hs.446466
          Hs.446466
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s230691

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WASH3P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003659.2 3 1059
          GenBank mRNA AK056232.1 - 3 900
          AK057951.1 - 2 255
          AK095183.1 - 3 253
          AK290087.1 - 2 258
          AK291582.1 - 3 268
          AK295256.1 - 3 265
          AK300161.1 - 4 481
          BC048328.1 - 2 255
          BC063555.1 - 3 457
          BC110996.1 - 3 757
          BM839085.1 - 2 254
          BQ722707.1 - 2 269
          BX452679.2 - 392
          Gene Symbol: LOC100288778
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_130745.1 3 381
          GenBank mRNA CN404396.1 - 2 245
          DA015248.1 - 3 267
          DC319181.1 - 3 384
          Gene Symbol: WASH1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_182905.4 NP_878908.4 3 381
          XM_011517658.2 XP_011515960.1 3 498
          XM_011517659.2 XP_011515961.1 3 280
          XM_011517660.2 XP_011515962.1 3 277
          XM_011517662.2 XP_011515964.1 3 1784
          XM_011517663.2 XP_011515965.1 2 1633
          XM_011517664.2 XP_011515966.1 2 1630
          XM_011517665.2 XP_011515967.1 2 179
          XM_011517671.2 XP_011515973.2 3 498
          XM_017014169.1 XP_016869658.1 3 397
          XM_017014170.1 XP_016869659.1 3 460
          XM_017014173.1 XP_016869662.1 3 498
          GenBank mRNA AK024481.1 - 1 443
          AK289708.1 - 3 384
          AK309568.1 - 2 113
          Gene Symbol: WASH2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024077.2 3 506
          GenBank mRNA AK292759.1 - 2 100
          AK308574.1 - 2 115
          AL137714.1 - 3 393
          AY217347.1 - 2 938
          BC082240.1 - 1 963
          BX377099.2 - 3 271
          BX537637.1 - 3 507
          Gene Symbol: WASH7P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024540.1 3 280
          GenBank mRNA AK294377.1 - 2 176
          AK296182.1 - 4 416
          AK302089.1 - 3 325
          AK308540.1 - 5 535
          AK309926.1 - 3 267
          AK310121.1 - 3 254
          AK310139.1 - 4 308
          AK316487.1 - 2 469
          BC094698.1 - 1 818
          DA054875.1 - 3 288
          DA143319.1 - 3 293

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to lysosome
          endosome organization
          endosomal transport
          Arp2/3 complex-mediated actin nucleation
          retrograde transport, endosome to Golgi
          early endosome to late endosome transport
          exocytosis
          protein transport
          extracellular matrix disassembly
          positive regulation of cell migration
          positive regulation of pseudopodium assembly
          retrograde transport, endosome to plasma membrane
          actin binding
          alpha-tubulin binding
          protein binding
          ubiquitin protein ligase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0