Search
Search

ATP10B| Gene Symbol: ATP10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025153.2 | NP_079429.2 | 26 | 5136 | ||
| XM_006714833.2 | XP_006714896.1 | 25 | 4717 | |||
| XM_011534468.2 | XP_011532770.1 | 25 | 4693 | |||
| XM_011534469.1 | XP_011532771.1 | 26 | 4957 | |||
| XM_011534472.2 | XP_011532774.1 | 21 | 4433 | |||
| XM_017009252.1 | XP_016864741.1 | 25 | 4657 | |||
| XM_017009253.1 | XP_016864742.1 | 24 | 4516 | |||
| GenBank mRNA | AB018258.2 | - | 26 | 4963 | ||
| AK025290.1 | - | 11 | 2059 | |||
| AK025294.1 | - | 8 | 1375 | |||