Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DEFA1 SIRNA Assays ›
          Gene Symbol:
          Defa1 Defa-rs7 Defa17 Defa23 Defa3 Gm15284 Gm15299
          Gene Name:
          defensin, alpha 1
          defensin, alpha, related sequence 7
          defensin, alpha, 17
          defensin, alpha, 23
          defensin, alpha, 3
          predicted gene 15284
          predicted pseudogene 15299
          Gene Aliases:
          Defcr, Defcr1
          CRS4C-2, CRS4C2, CRS4C2b, CRS4C2c, CRS4C2d, CRS4C3a, CRS4C3a2, CRS4C3b, CRS4C3c, CRS4C3d, CRS4C3e, Defcr-rs7
          AU014719, Cryp17, Defcr17
          Defcr23
          Defa27, Defcr-3, Defcr3
          OTTMUSG00000019742
          ENSMUSG00000074438, OTTMUSG00000019793
          Chromosome Location:
          -
          Chromosome Location:
          Chr.8: 21191573 - 21192550 on Build GRCm38
          Chromosome Location:
          Chr.8: 21655767 - 21656736 on Build GRCm38
          Chromosome Location:
          Chr.8: 21055040 - 21056016 on Build GRCm38
          Chromosome Location:
          Chr.8: 21287409 - 21288377 on Build GRCm38
          Chromosome Location:
          Chr.8: 21134650 - 21135598 on Build GRCm38
          Chromosome Location:
          Chr.8: 21315546 - 21316387 on Build GRCm38
          UniGene ID:
          Mm.425522
          Mm.442051
          Mm.425519
          Mm.425522
          Mm.436573
          Mm.426552
          Mm.479864
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s232275

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Defa1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010031.2 NP_034161.2 1, 2 217
          GenBank mRNA M33225.1 - 1, 2 217
          X15617.1 - 1, 2 217
          Gene Symbol: Defa-rs7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007848.2 NP_031874.2 1, 2 214
          Gene Symbol: Defa17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167790.1 NP_001161262.1 1, 2 207
          Gene Symbol: Defa23
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012307.2 NP_001012307.1 1, 2 214
          GenBank mRNA AY746428.1 - 1, 2 173
          BC125548.1 - 1, 2 194 40130063
          BC125550.1 - 1, 2 194 40130065
          Gene Symbol: Defa3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007850.2 NP_031876.1 1, 2 207
          GenBank mRNA BC099854.1 - 1, 2 168
          BC099855.1 - 1, 2 168
          S73391.1 - 187
          Gene Symbol: Gm15284
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177485.1 NP_001170956.1 1, 2 186
          Gene Symbol: Gm15299
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170955.1 NP_001164426.1 1, 2 173

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          antimicrobial response protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          response to drug
          defense response to bacterium
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          cellular response to lipopolysaccharide
          biological_process
          protein homodimerization activity
          protein heterodimerization activity
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0