Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIST1H3F SIRNA Assays ›
          Gene Symbol:
          Hist1h3f Hist1h3a Hist1h3b Hist1h3c Hist1h3d Hist1h3e Hist1h3g Hist1h3h Hist1h3i Hist2h3b Hist2h3c1 Hist2h3c2
          Gene Name:
          histone cluster 1, H3f
          histone cluster 1, H3a
          histone cluster 1, H3b
          histone cluster 1, H3c
          histone cluster 1, H3d
          histone cluster 1, H3e
          histone cluster 1, H3g
          histone cluster 1, H3h
          histone cluster 1, H3i
          histone cluster 2, H3b
          histone cluster 2, H3c1
          histone cluster 2, H3c2
          Gene Aliases:
          H3.2-221, Hist1
          -
          H3-53
          H3-143
          H3-b
          H3-f, H3.1
          H3.1-221, M32460
          H3-291
          H3-H
          H3-615, H3-616
          BE691662, H3-615, H3f2, Hist2h3ca1
          H3-614, H3.2-614, Hist2h3c2-ps, Hist2h3ca1, M32461
          Chromosome Location:
          Chr.13: 23544052 - 23544954 on Build GRCm38
          Chromosome Location:
          Chr.13: 23761885 - 23762386 on Build GRCm38
          Chromosome Location:
          Chr.13: 23752380 - 23752840 on Build GRCm38
          Chromosome Location:
          Chr.13: 23745042 - 23745521 on Build GRCm38
          Chromosome Location:
          Chr.13: 23575763 - 23576266 on Build GRCm38
          Chromosome Location:
          Chr.13: 23561896 - 23562365 on Build GRCm38
          Chromosome Location:
          Chr.13: 23535418 - 23535909 on Build GRCm38
          Chromosome Location:
          Chr.13: 21717628 - 21718115 on Build GRCm38
          Chromosome Location:
          Chr.13: 21782915 - 21783397 on Build GRCm38
          Chromosome Location:
          Chr.3: 96268654 - 96269155 on Build GRCm38
          Chromosome Location:
          Chr.3: 96246685 - 96247348 on Build GRCm38
          Chromosome Location:
          Chr.3: 96238479 - 96239127 on Build GRCm38
          UniGene ID:
          Mm.221389
          Mm.221301
          -
          Mm.262517
          Mm.347699
          Mm.261658
          Mm.261657
          Mm.377874
          Mm.390558
          Mm.383293
          Mm.422680
          Mm.358797
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s233207

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hist1h3f
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013548.4 NP_038576.1 1 662
          GenBank mRNA AK010121.1 - 1 251
          BC101954.1 - 1 662 40044770
          BC101955.1 - 1 662 40044769
          BC101956.1 - 1 662 40044765
          BC103549.1 - 1 662 40044771
          Gene Symbol: Hist1h3a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013550.5 NP_038578.2 1 262
          GenBank mRNA AK006742.1 - 1 242
          AK018952.1 - 1 263
          BC115816.1 - 1 259 40105082
          Gene Symbol: Hist1h3b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178203.2 NP_835510.1 1 229
          Gene Symbol: Hist1h3c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175653.2 NP_783584.1 1 253
          GenBank mRNA BC120800.1 - 1 334 40129723
          BC120802.1 - 1 334 40129725
          Gene Symbol: Hist1h3d
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178204.2 NP_835511.1 1 268
          GenBank mRNA AK020421.1 - 269
          Gene Symbol: Hist1h3e
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178205.2 NP_835512.1 1 229
          GenBank mRNA BC127042.1 - 1 222
          Gene Symbol: Hist1h3g
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_145073.2 NP_659539.1 1 257
          GenBank mRNA BC125355.1 - 1 253 40129870
          BC125357.1 - 1 253 40129872
          Gene Symbol: Hist1h3h
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178206.2 NP_835513.1 1 260
          GenBank mRNA BC107285.2 - 1 227
          Gene Symbol: Hist1h3i
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178207.2 NP_835514.1 1 256
          GenBank mRNA AK155722.1 - 257
          BC107287.2 - 1 225
          BC116383.2 - 1 229 40101629
          BC127148.1 - 1 228
          Gene Symbol: Hist2h3b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178215.2 NP_835587.1 1 269
          GenBank mRNA BC139251.1 - 1 269 8862273
          BC139252.1 - 1 269 8862274
          Gene Symbol: Hist2h3c1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178216.3 NP_835734.2 1 430
          GenBank mRNA BC094041.1 - 430 5099464
          Gene Symbol: Hist2h3c2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_054045.4 NP_473386.1 1 415
          GenBank mRNA BC015270.1 - 1 415 4240748
          BC132488.1 - 1 259 40130765
          BC132490.1 - 1 259 40130767

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001107698, XM_008761309, XM_008761346, XM_008771686, XM_001057443.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          positive regulation of defense response to virus by host
          DNA replication-dependent nucleosome assembly
          protein heterotetramerization
          regulation of gene silencing
          negative regulation of transcription from RNA polymerase II promoter
          histone binding
          chromatin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0