Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SERPINA1D SIRNA Assays ›
          Gene Symbol:
          Serpina1d Serpina1a Serpina1b Serpina1c Serpina1e
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade A, member 1D
          serine (or cysteine) peptidase inhibitor, clade A, member 1A
          serine (or cysteine) preptidase inhibitor, clade A, member 1B
          serine (or cysteine) peptidase inhibitor, clade A, member 1C
          serine (or cysteine) peptidase inhibitor, clade A, member 1E
          Gene Aliases:
          AI118301, Dom4, PI4, Spi1-4
          Aat-2, Aat2, Dom1, PI1, Spi1-1, Spi1-3
          D12Ucla2, Dom2, PI2, Spi1-2
          Pi3, Pi6, Skalp, Spi1-3, Spi1-6, Wap3
          Dom5, PI5, Spi1-5
          Chromosome Location:
          Chr.12: 103763587 - 103773633 on Build GRCm38
          Chromosome Location:
          Chr.12: 103853295 - 103863619 on Build GRCm38
          Chromosome Location:
          Chr.12: 103728156 - 103738189 on Build GRCm38
          Chromosome Location:
          Chr.12: 103894926 - 103904950 on Build GRCm38
          Chromosome Location:
          Chr.12: 103946931 - 103956897 on Build GRCm38
          UniGene ID:
          Mm.439695
          Mm.439692
          Mm.439692
          Mm.439692
          Mm.312593
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s233445

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Serpina1d
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009246.3 NP_033272.1 5 1186
          GenBank mRNA AK002537.1 - 5 1146
          BC021850.1 - 4 1122 5125194
          M75718.1 - 5 1127
          Gene Symbol: Serpina1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252569.1 NP_001239498.1 6 1396
          NM_009243.4 NP_033269.1 5 1181
          GenBank mRNA AK146619.1 - 1114
          BC011040.1 - 1107 4159448
          BC037007.1 - 1333 5123904
          BC047128.1 - 1107
          BC049970.1 - 1107 4160332
          BC057982.1 - 1114 5134754
          BC057984.1 - 1129 5126441
          BC057989.1 - 1092 5100593
          M75721.1 - 1112
          W82055.1 - 2 110
          Gene Symbol: Serpina1b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009244.4 NP_033270.3 5 1176
          GenBank mRNA BC012874.1 - 1117 4194027
          BC022109.1 - 1105 5133541
          BC025445.1 - 1116 5053771
          BC037008.1 - 1111
          BC049255.1 - 1327 5137448
          BC106098.1 - 1511 4223129
          M25529.1 - 1087
          M75716.1 - 4 1046
          Gene Symbol: Serpina1c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC009818.1 - 1118 4211464
          BC010984.1 - 1118 4163602
          BC010988.1 - 1116
          BC011041.1 - 1119 4193647
          BC015266.1 - 1070
          BC021325.1 - 1159 5052898
          BC021780.1 - 1138 4160260
          BC024108.1 - 1129 5099206
          BC031707.1 - 1105 5123976
          Gene Symbol: Serpina1e
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009247.2 NP_033273.1 5 1124
          GenBank mRNA AK004999.1 - 5 1126
          AK146576.1 - 1326
          BC061176.1 - 5 1135 30381238
          M75717.1 - 5 1119

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          negative regulation of endopeptidase activity
          response to cytokine
          response to peptide hormone
          protein N-linked glycosylation
          acute-phase response
          in utero embryonic development
          glycoprotein binding
          protease binding
          endopeptidase inhibitor activity
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity
          identical protein binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0