Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DEFA14 SIRNA Assays ›
          Gene Symbol:
          Defa14 Defa-rs7 Defa12 Defa16 Defa17 Defa2 Defa23 Defa24 Defa3 Defa6 Gm15284
          Gene Name:
          defensin, alpha, 14
          defensin, alpha, related sequence 7
          defensin, alpha, 12
          defensin, alpha, 16
          defensin, alpha, 17
          defensin, alpha, 2
          defensin, alpha, 23
          defensin, alpha, 24
          defensin, alpha, 3
          defensin, alpha, 6
          predicted gene 15284
          Gene Aliases:
          Defcr14
          CRS4C-2, CRS4C2, CRS4C2b, CRS4C2c, CRS4C2d, CRS4C3a, CRS4C3a2, CRS4C3b, CRS4C3c, CRS4C3d, CRS4C3e, Defcr-rs7
          Defcr12
          Defcr16
          AU014719, Cryp17, Defcr17
          CR2, Defcr-2, Defcr2
          Defcr23
          Defcr24
          Defa27, Defcr-3, Defcr3
          Defcr6
          OTTMUSG00000019742
          Chromosome Location:
          -
          Chromosome Location:
          Chr.8: 21191573 - 21192550 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.8: 21655767 - 21656736 on Build GRCm38
          Chromosome Location:
          Chr.8: 21378517 - 21379495 on Build GRCm38
          Chromosome Location:
          Chr.8: 21055040 - 21056016 on Build GRCm38
          Chromosome Location:
          Chr.8: 21734494 - 21735471 on Build GRCm38
          Chromosome Location:
          Chr.8: 21287409 - 21288377 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.8: 21134650 - 21135598 on Build GRCm38
          UniGene ID:
          Mm.425522
          Mm.442051
          Mm.477846
          Mm.425522
          Mm.425519
          Mm.327348
          Mm.425522
          Mm.443396
          Mm.436573
          Mm.426552
          Mm.426552
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s233787

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Defa14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U03067.1 - 1 56
          Gene Symbol: Defa-rs7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007848.2 NP_031874.2 2 269
          Gene Symbol: Defa12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U03063.1 - 1 56
          Gene Symbol: Defa16
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U03066.1 - 1 56
          Gene Symbol: Defa17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167790.1 NP_001161262.1 2 262
          Gene Symbol: Defa2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U03028.1 - 56
          Gene Symbol: Defa23
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012307.2 NP_001012307.1 2 269
          GenBank mRNA AV052259.1 - 2 148
          AY746428.1 - 2 228
          BC125548.1 - 2 249 40130063
          BC125550.1 - 2 249 40130065
          Gene Symbol: Defa24
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024225.2 NP_001019396.1 2 271
          GenBank mRNA AY746429.1 - 2 228
          Gene Symbol: Defa3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007850.2 NP_031876.1 2 262
          GenBank mRNA BC099854.1 - 2 223
          BC099855.1 - 2 223
          U03030.1 - 56
          Gene Symbol: Defa6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007852.1 NP_031878.1 2 228
          GenBank mRNA U03034.1 - 1 56
          Gene Symbol: Gm15284
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177485.1 NP_001170956.1 2 241

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          antimicrobial response protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response to bacterium
          biological_process
          defense response
          antibacterial humoral response
          response to drug
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          cellular response to lipopolysaccharide
          protein homodimerization activity
          protein heterodimerization activity
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0