Search
Search

Ssxb2| Gene Symbol: Ssxb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001450.4 | NP_001001450.1 | 5 | 561 | ||
| NM_001134226.1 | NP_001127698.1 | 4 | 384 | |||
| GenBank mRNA | AK076879.1 | - | 5 | 562 | ||
| AY244793.1 | - | 5 | 562 | |||
| BC048441.1 | - | 4 | 384 | 6773922 | ||
| BC089478.1 | - | 4 | 384 | 6743842 | ||