Search
Search

Cnot2| Gene Symbol: Cnot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037846.3 | NP_001032935.2 | 16 | 2165 | ||
| NM_001037847.2 | NP_001032936.2 | 16 | 2095 | |||
| NM_028082.2 | NP_082358.2 | 15 | 2042 | |||
| GenBank mRNA | AK011231.1 | - | 14 | 1965 | ||
| AK078121.1 | - | 16 | 2155 | |||
| AK149767.1 | - | 16 | 2139 | |||
| AK167834.1 | - | 16 | 2144 | |||
| BC043133.1 | - | 16 | 2066 | |||
| BC063105.1 | - | 17 | 2308 | 5003026 | ||
| BC065171.1 | - | 16 | 2095 | 6415034 | ||
| BC090624.1 | - | 14 | 1971 | 6815664 | ||