Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SSXB10 SIRNA Assays ›
          Gene Symbol:
          Ssxb10 Gm14459 Gm6592 Ssx9 Ssxb1 Ssxb11 Ssxb12 Ssxb3 Ssxb5 Ssxb6
          Gene Name:
          synovial sarcoma, X member B, breakpoint 10
          predicted gene 14459
          predicted gene 6592
          synovial sarcoma, X breakpoint 9
          synovial sarcoma, X member B, breakpoint 1
          synovial sarcoma, X member B, breakpoint 11
          synovial sarcoma, X member B, breakpoint 12
          synovial sarcoma, X member B, breakpoint 3
          synovial sarcoma, X member B, breakpoint 5
          synovial sarcoma, X member B, breakpoint 6
          Gene Aliases:
          -
          OTTMUSG00000016698
          EG625480
          Ssxb4
          4930414C09Rik
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.X: 8327424 - 8336235 on Build GRCm38
          Chromosome Location:
          Chr.X: 8513543 - 8526726 on Build GRCm38
          Chromosome Location:
          Chr.X: 8843994 - 8849607 on Build GRCm38
          Chromosome Location:
          Chr.X: 8743792 - 8756633 on Build GRCm38
          Chromosome Location:
          Chr.X: 8413305 - 8422157 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.X: 8583404 - 8591810 on Build GRCm38
          Chromosome Location:
          Chr.X: 8803691 - 8809445 on Build GRCm38
          Chromosome Location:
          Chr.X: 8542179 - 8548362 on Build GRCm38
          UniGene ID:
          Mm.484393
          Mm.475615
          Mm.430078
          Mm.391220
          Mm.298030
          Mm.298030
          Mm.298030
          Mm.379565
          Mm.227643
          Mm.227643
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234318

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ssxb10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199064.1 NP_951019.1 7 647
          GenBank mRNA AY244801.1 - 7 647
          BC145539.1 - 619 9054102
          Gene Symbol: Gm14459
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001126491.1 NP_001119963.1 6 492
          Gene Symbol: Gm6592
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081564.1 NP_001075033.1 6 459
          Gene Symbol: Ssx9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199063.2 NP_951018.1 6 511
          GenBank mRNA AY244795.1 - 6 511
          Gene Symbol: Ssxb1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_026492.3 NP_080768.1 7 646
          GenBank mRNA AK006218.1 - 7 647
          AK015135.1 - 7 647
          AY244792.1 - 7 647
          BC145538.1 - 7 619 9054101
          BC145541.1 - 564 9054104
          BC145782.1 - 7 538 40131085
          BC145784.1 - 7 538 40131087
          BC171964.1 - 619 9054250
          Gene Symbol: Ssxb11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY244802.1 - 6 510
          Gene Symbol: Ssxb12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY244803.1 - 6 510
          Gene Symbol: Ssxb3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_198898.2 NP_942601.1 6 511
          GenBank mRNA AY244794.1 - 6 511
          BC087939.1 - 7 591
          BC132618.1 - 6 511 40130895
          BC132644.1 - 6 511 40130921
          Gene Symbol: Ssxb5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199319.3 NP_955023.1 6 510
          GenBank mRNA AY244796.1 - 6 510
          Gene Symbol: Ssxb6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001205108.1 NP_001192037.1 470

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0