Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS2 SIRNA Assays ›
          Gene Symbol:
          Rps2 Gm10653 Rps2-ps6
          Gene Name:
          ribosomal protein S2
          ribsomal protein S2 pseudogene
          ribosomal protein S2, pseudogene 6
          Gene Aliases:
          Llrep3, S4
          ENSMUSG00000074250
          EG628962, Gm16408
          Chromosome Location:
          Chr.17: 24718121 - 24721927 on Build GRCm38
          Chromosome Location:
          Chr.9: 62841477 - 62862024 on Build GRCm38
          Chromosome Location:
          Chr.14: 76236647 - 76237574 on Build GRCm38
          UniGene ID:
          Mm.157452
          Mm.422004
          Mm.308628
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s234418

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008503.5 NP_032529.2 5, 6 867
          GenBank mRNA AF283559.1 - 4, 5 794
          AK004568.1 - 647
          AK012360.1 - 5, 6 823
          AK019265.1 - 5, 6 823
          AK146259.1 - 5, 6 822
          AK151985.1 - 5, 6 821
          AK152299.1 - 5, 6 821
          AK159121.1 - 5, 6 821
          AK159138.1 - 5, 6 819
          AK163821.1 - 4, 5 961
          AK166549.1 - 5, 6 821
          AK166664.1 - 5, 6 824
          AK166726.1 - 5, 6 821
          AK167033.1 - 5, 6 820
          AK167799.1 - 5, 6 821
          AK167974.1 - 5, 6 821
          AK168307.1 - 5, 6 820
          AK168528.1 - 5, 6 821
          AK168630.1 - 5, 6 821
          AK169119.1 - 5, 6 821
          AK190542.1 - 2, 3 144
          AK191166.1 - 2, 3 144
          AK191286.1 - 2, 3 387
          AK191883.1 - 144
          AK193261.1 - 2, 3 145
          AK196057.1 - 2, 3 144
          AK197334.1 - 2, 3 144
          AK197696.1 - 2, 3 144
          AK205379.1 - 144
          AK215035.1 - 2, 3 144
          BC002186.1 - 812 3487739
          BC087956.1 - 5, 6 823 6749924
          BC091730.1 - 4, 5 798 2651289
          BC091755.1 - 4, 5 805 4220412
          BC092286.1 - 4, 5 800 5007960
          BC093493.1 - 4, 5 796 2655743
          BC094058.1 - 4, 5 797 3675994
          BC094273.1 - 4, 5 799 2654868
          BC106136.1 - 4, 5 797 3676057
          BC110678.1 - 4, 5 805 4011338
          BC116446.1 - 4, 5 797 3496118
          BC117558.1 - 4, 5 795 5034223
          BY443672.1 - 2, 3 293
          U89418.1 - 3 519
          Gene Symbol: Gm10653
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK131930.1 - 1124
          Gene Symbol: Rps2-ps6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK186621.1 - 1 144
          AK188199.1 - 1 144

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031838, FQ222558, FQ229778, U92696, FQ218643, and others.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          formation of translation preinitiation complex
          translation
          positive regulation of transferase activity
          cellular response to interleukin-4
          RNA binding
          mRNA binding
          structural constituent of ribosome
          fibroblast growth factor binding
          enzyme binding
          protein complex binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline