Search
Search

| Gene Symbol: Rbpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106631.1 | NP_001100101.1 | 6 | 469 | ||
| XM_008770190.1 | XP_008768412.1 | 5 | 1140 | |||
| XM_008770191.1 | XP_008768413.1 | 5 | 381 | |||
| XM_008770192.1 | XP_008768414.1 | 5 | 2559 | |||
| XM_008770193.1 | XP_008768415.1 | 6 | 552 | |||
| Gene Symbol: Rbpjl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237737.2 | XP_006237799.1 | 2 | 391 | ||