Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RGD1565355 SIRNA Assays ›
          Gene Symbol:
          RGD1565355 Cd36 LOC103690020
          Gene Name:
          similar to fatty acid translocase/CD36
          CD36 molecule (thrombospondin receptor)
          platelet glycoprotein 4-like
          Gene Aliases:
          -
          Fat
          -
          Chromosome Location:
          Chr.4: 14071705 - 14112330 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 14150309 - 14191498 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 14205809 - 14247610 on Build Rnor_6.0
          UniGene ID:
          Rn.225150
          Rn.102418
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s235391

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RGD1565355
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001109218.1 NP_001102688.1 136
          GenBank mRNA BC091104.1 - 334 7383442
          Gene Symbol: Cd36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031561.2 NP_113749.2 345
          GenBank mRNA AB005743.1 - 407
          AF072411.1 - 1, 2 136
          AF111268.1 - 152
          AF113914.1 - 1, 2 130
          BC072543.1 - 345 7105943
          L19658.1 - 1, 2 185
          Gene Symbol: LOC103690020
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006235962.2 XP_006236024.1 3, 4 771
          XM_006235963.2 XP_006236025.1 344
          XM_006235964.2 XP_006236026.1 3, 4 787
          XM_006235965.2 XP_006236027.1 3, 4 776

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001159556, NM_001159557, NM_007643, NM_001159558, NM_001159555, and others.

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          negative regulation of transcription from RNA polymerase II promoter
          long-chain fatty acid metabolic process
          positive regulation of cell-matrix adhesion
          pattern recognition receptor signaling pathway
          negative regulation of systemic arterial blood pressure
          fatty acid metabolic process
          triglyceride metabolic process
          receptor-mediated endocytosis
          phagocytosis, recognition
          signal transduction
          cell surface receptor signaling pathway
          nitric oxide mediated signal transduction
          response to nutrient
          response to mechanical stimulus
          negative regulation of gene expression
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cholesterol storage
          response to activity
          fatty acid transport
          long-chain fatty acid transport
          plasma membrane long-chain fatty acid transport
          negative regulation of angiogenesis
          fatty acid oxidation
          lipid storage
          cGMP-mediated signaling
          positive regulation of blood coagulation
          cholesterol transport
          response to estradiol
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          cellular response to insulin stimulus
          plasma lipoprotein particle clearance
          low-density lipoprotein particle clearance
          response to drug
          lipoprotein transport
          negative regulation of transcription factor import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          apoptotic cell clearance
          positive regulation of MAPK cascade
          negative regulation of growth of symbiont in host
          long-chain fatty acid import
          digestive tract development
          positive regulation of cytokine secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-positive bacterium
          low-density lipoprotein particle mediated signaling
          positive regulation of phagocytosis, engulfment
          response to growth hormone
          positive regulation of macrophage cytokine production
          cellular response to bacterial lipopeptide
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          cellular response to hydroperoxide
          regulation of removal of superoxide radicals
          positive regulation of blood microparticle formation
          positive regulation of reactive oxygen species metabolic process
          low-density lipoprotein receptor activity
          protein binding
          high-density lipoprotein particle binding
          lipid binding
          low-density lipoprotein particle binding
          transforming growth factor beta binding
          thrombospondin receptor activity
          oleic acid binding
          lipoteichoic acid receptor activity
          lipoprotein particle binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0