Search
Search

CAMK2A| Gene Symbol: CAMK2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015981.3 | NP_057065.2 | 16 | 1346 | ||
| NM_171825.2 | NP_741960.1 | 15 | 1313 | |||
| XM_017009898.1 | XP_016865387.1 | 15 | 1313 | |||
| GenBank mRNA | AB023185.1 | - | 15 | 1246 | ||
| AF091486.1 | - | 15 | 1202 | |||
| AK289776.1 | - | 16 | 1346 | |||
| BC040457.1 | - | 15 | 1228 | 4811948 | ||