Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DICER1 SIRNA Assays ›
          Gene Symbol:
          DICER1
          Gene Name:
          dicer 1, ribonuclease III
          Gene Aliases:
          DCR1, Dicer, Dicer1e, HERNA, K12H4.8-LIKE, MNG1, RMSE2
          Chromosome Location:
          Chr.14: 95086228 - 95158010 on Build GRCh38
          UniGene ID:
          Hs.738957
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s23756
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DICER1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195573.1 NP_001182502.1 8 1466
          NM_001271282.2 NP_001258211.1 9 1654
          NM_001291628.1 NP_001278557.1 9 1542
          NM_030621.4 NP_085124.2 11 1713
          NM_177438.2 NP_803187.1 9 1659
          XM_011536599.1 XP_011534901.1 11 1845
          XM_011536600.2 XP_011534902.1 11 1720
          XM_011536601.2 XP_011534903.1 12 1865
          XM_011536602.2 XP_011534904.1 12 1756
          XM_011536604.2 XP_011534906.1 8 1541
          XM_011536605.2 XP_011534907.1 2 6346
          XM_017021120.1 XP_016876609.1 11 2152
          XM_017021121.1 XP_016876610.1 9 1547
          XM_017021122.1 XP_016876611.1 10 1591
          XM_017021123.1 XP_016876612.1 10 2022
          GenBank mRNA AB023145.2 - 9 1659
          AB028449.1 - 10 1609
          AJ132261.2 - 9 1662
          BC150287.1 - 9 1659 8860125
          HM595745.1 - 8 1430

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00229023_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          apoptotic DNA fragmentation
          negative regulation of Schwann cell proliferation
          positive regulation of Schwann cell differentiation
          nerve development
          production of siRNA involved in RNA interference
          targeting of mRNA for destruction involved in RNA interference
          gene silencing by RNA
          pre-miRNA processing
          positive regulation of myelination
          peripheral nervous system myelin formation
          conversion of ds siRNA to ss siRNA involved in RNA interference
          siRNA loading onto RISC involved in RNA interference
          production of miRNAs involved in gene silencing by miRNA
          miRNA loading onto RISC involved in gene silencing by miRNA
          conversion of ds siRNA to ss siRNA
          neuron projection morphogenesis
          RNA phosphodiester bond hydrolysis, endonucleolytic
          double-stranded RNA binding
          helicase activity
          endoribonuclease activity
          ribonuclease III activity
          deoxyribonuclease I activity
          protein binding
          ATP binding
          protein domain specific binding
          siRNA binding
          metal ion binding
          pre-miRNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0