Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HEY1 SIRNA Assays ›
          Gene Symbol:
          HEY1
          Gene Name:
          hes related family bHLH transcription factor with YRPW motif 1
          Gene Aliases:
          BHLHb31, CHF2, HERP2, HESR1, HRT-1, OAF1, hHRT1
          Chromosome Location:
          Chr.8: 79764010 - 79767863 on Build GRCh38
          UniGene ID:
          Hs.234434
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s23868

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HEY1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040708.1 NP_001035798.1 5 636
          NM_001282851.1 NP_001269780.1 2 366
          NM_012258.3 NP_036390.3 5 624
          GenBank mRNA AF151522.1 - 5 448
          AF176422.1 - 5 525
          AF232239.1 - 5 530
          AF254637.1 - 5 541
          AF311883.1 - 5 424
          AJ272214.1 - 5 509
          AK074641.1 - 5 527
          AK092437.1 - 5 543
          AK226041.1 - 5 531
          AK293646.1 - 2 366
          AK307622.1 - 2 220
          AK313271.1 - 5 528
          BC001873.1 - 5 521 3533622
          BP283540.1 - 5 573
          BT020065.1 - 5 424

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          pulmonary valve morphogenesis
          atrioventricular valve formation
          epithelial to mesenchymal transition involved in endocardial cushion formation
          endocardial cushion morphogenesis
          cardiac ventricle morphogenesis
          transcription, DNA-templated
          Notch signaling pathway
          dorsal aorta morphogenesis
          umbilical cord morphogenesis
          negative regulation of Notch signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cardiac epithelial to mesenchymal transition
          heart trabecula formation
          cardiac septum morphogenesis
          ventricular septum morphogenesis
          labyrinthine layer blood vessel development
          arterial endothelial cell differentiation
          Notch signaling involved in heart development
          cellular response to glucocorticoid stimulus
          negative regulation of transcription regulatory region DNA binding
          negative regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
          regulation of vasculogenesis
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          transcription factor activity, protein binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          microsatellite binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0