Search
Search

SUN2| Gene Symbol: SUN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199579.1 | NP_001186508.1 | 16 | 2239 | ||
| NM_001199580.1 | NP_001186509.1 | 17 | 2175 | |||
| NM_015374.2 | NP_056189.1 | 16 | 2176 | |||
| XM_011530104.2 | XP_011528406.1 | 15 | 2033 | |||
| XM_011530105.2 | XP_011528407.1 | 16 | 1900 | |||
| XM_017028748.1 | XP_016884237.1 | 16 | 2171 | |||
| GenBank mRNA | AB014568.1 | - | 16 | 1958 | ||
| AF202723.1 | - | 8 | 1051 | |||
| AK295787.1 | - | 16 | 1900 | |||
| AK304188.1 | - | 17 | 2035 | |||
| AY682988.1 | - | 16 | 1957 | |||
| BC030684.2 | - | 4 | 381 | |||
| BC069253.1 | - | 8 | 1057 | |||
| BC094797.1 | - | 16 | 2196 | 4799090 | ||
| BC111549.1 | - | 15 | 1905 | 40008751 | ||
| BC111717.1 | - | 15 | 1905 | 40008756 | ||
| BX537962.1 | - | 17 | 2167 | |||
| CR456474.1 | - | 15 | 1903 | |||