Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CIZ1 SIRNA Assays ›
          Gene Symbol:
          CIZ1
          Gene Name:
          CDKN1A interacting zinc finger protein 1
          Gene Aliases:
          LSFR1, NP94, ZNF356
          Chromosome Location:
          Chr.9: 128166065 - 128204383 on Build GRCh38
          UniGene ID:
          Hs.212395
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s24489

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CIZ1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001131015.1 NP_001124487.1 14, 15 2191
          NM_001131016.1 NP_001124488.1 13, 14 2319
          NM_001131017.1 NP_001124489.1 14, 15 2136
          NM_001131018.1 NP_001124490.1 13, 14 2079
          NM_001257975.1 NP_001244904.1 14, 15 2489
          NM_001257976.1 NP_001244905.1 12, 13 2203
          NM_012127.2 NP_036259.2 13, 14 2359
          XM_005251888.3 XP_005251945.2 13, 14 2190
          XM_005251891.3 XP_005251948.2 14, 15 2022
          XM_005251892.3 XP_005251949.2 14, 15 2007
          XM_005251893.4 XP_005251950.4 13, 14 1945
          XM_006717039.3 XP_006717102.2 12, 13 2098
          XM_011518481.1 XP_011516783.1 13, 14 2175
          XM_011518482.2 XP_011516784.2 12, 13 2113
          XM_011518483.1 XP_011516785.1 12, 13 2230
          XM_017014594.1 XP_016870083.1 13, 14 2643
          XM_017014595.1 XP_016870084.1 13, 14 2287
          XM_017014596.1 XP_016870085.1 14, 15 2134
          GenBank mRNA AB030835.1 - 13, 14 2272
          AF159025.1 - 12, 13 2151
          AF234161.1 - 11, 12 1922
          AK023978.1 - 14, 15 2191
          AK292713.1 - 13, 14 2310
          AK296363.1 - 14, 15 2490
          AK303288.1 - 14, 15 2119
          AK303636.1 - 12, 13 2186
          AK316393.1 - 11, 12 1915
          BC004119.2 - 14, 15 1976 3507940
          BC021163.2 - 13, 14 2058 3633796
          EF441870.1 - 6, 7 865

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          protein binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0