Search
Search

FBXO7| Gene Symbol: FBXO7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033024.1 | NP_001028196.1 | 2, 3 | 298 | ||
| NM_001257990.1 | NP_001244919.1 | 2, 3 | 450 | |||
| NM_012179.3 | NP_036311.3 | 2, 3 | 693 | |||
| GenBank mRNA | AF129537.1 | - | 1, 2 | 307 | ||
| AF233225.1 | - | 2, 3 | 413 | |||
| AK226087.1 | - | 2, 3 | 608 | |||
| AK291972.1 | - | 2, 3 | 433 | |||
| AK297841.1 | - | 2, 3 | 253 | |||
| AK301716.1 | - | 2, 3 | 378 | |||
| AL050254.1 | - | 2, 3 | 615 | |||
| BC008361.1 | - | 2, 3 | 690 | 3611049 | ||
| BI821903.1 | - | 2, 3 | 298 | |||
| BX362053.2 | - | 2, 3 | 664 | |||
| BX648151.1 | - | 2, 3 | 246 | |||
| BX648220.1 | - | 2, 3 | 243 | |||
| CR456491.1 | - | 2, 3 | 528 | |||