Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CBLB SIRNA Assays ›
Gene Symbol:
CBLB
Gene Name:
Cbl proto-oncogene B
Gene Aliases:
Cbl-b, Nbla00127, RNF56
Chromosome Location:
Chr.3: 105655461 - 105869552 on Build GRCh38
UniGene ID:
Hs.430589
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2479
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CBLB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321786.1
NP_001308715.1
6, 7
1077
NM_001321788.1
NP_001308717.1
6, 7
996
NM_001321789.1
NP_001308718.1
6, 7
1015
NM_001321790.1
NP_001308719.1
6, 7
950
NM_001321791.1
NP_001308720.1
6, 7
996
NM_001321793.1
NP_001308722.1
6, 7
1172
NM_001321794.1
NP_001308723.1
6, 7
996
NM_001321795.1
NP_001308724.1
6, 7
1172
NM_001321796.1
NP_001308725.1
5, 6
849
NM_001321797.1
NP_001308726.1
6, 7
996
NM_001321798.1
NP_001308727.1
6, 7
1172
NM_001321799.1
NP_001308728.1
5, 6
1025
NM_001321806.1
NP_001308735.1
5, 6
839
NM_001321807.1
NP_001308736.1
7, 8
1316
NM_001321808.1
NP_001308737.1
7, 8
1140
NM_001321811.1
NP_001308740.1
5, 6
1015
NM_001321813.1
NP_001308742.1
7, 8
1053
NM_001321816.1
NP_001308745.1
5, 6
1015
NM_001321820.1
NP_001308749.1
7, 8
1140
NM_001321822.1
NP_001308751.1
6, 7
1172
NM_170662.4
NP_733762.2
6, 7
1172
NR_135806.1
7, 8
1304
NR_135807.1
7, 8
1220
NR_135808.1
6, 7
1172
NR_135809.1
6, 7
1172
NR_135810.1
6, 7
950
NR_135811.1
6, 7
996
NR_135812.1
6, 7
892
XM_011513257.1
XP_011511559.1
6, 7
992
XM_011513259.2
XP_011511561.2
6, 7
1117
XM_017007395.1
XP_016862884.1
6, 7
944
XM_017007396.1
XP_016862885.1
6, 7
1115
XM_017007397.1
XP_016862886.1
5, 6
969
XM_017007398.1
XP_016862887.1
5, 6
795
XM_017007399.1
XP_016862888.1
5, 6
1020
XM_017007400.1
XP_016862889.1
5, 6
969
GenBank mRNA
AK292792.1
-
6, 7
1113
AK302533.1
-
6, 7
950
AK302892.1
-
6, 7
925
AK316276.1
-
6, 7
930
BC032851.2
-
6, 7
1113
5269036
DQ349203.1
-
5, 6
850
DQ349204.1
-
5, 6
850
U26710.1
-
6, 7
1172
U26711.1
-
6, 7
1172
U26712.1
-
6, 7
1172
Back To Top
More Information
Panther Classification:
Molecular Function -
ligase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
NLS-bearing protein import into nucleus
signal transduction
cell surface receptor signaling pathway
response to gravity
response to hormone
regulation of signaling
phosphotyrosine binding
ubiquitin-protein transferase activity
signal transducer activity
calcium ion binding
protein binding
zinc ion binding
ligase activity
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0