Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIPA1L1 SIRNA Assays ›
Gene Symbol:
SIPA1L1
Gene Name:
signal induced proliferation associated 1 like 1
Gene Aliases:
E6TP1
Chromosome Location:
Chr.14: 71320418 - 71741229 on Build GRCh38
UniGene ID:
Hs.654657
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24985
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIPA1L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284245.1
NP_001271174.1
17
5207
NM_001284246.1
NP_001271175.1
17
5796
NM_001284247.1
NP_001271176.1
17
5211
NM_015556.2
NP_056371.1
18
5270
XM_005267516.4
XP_005267573.1
18
5859
XM_006720111.2
XP_006720174.1
20
7149
XM_011536630.1
XP_011534932.1
22
5621
XM_011536631.1
XP_011534933.1
22
6032
XM_011536632.1
XP_011534934.1
21
5541
XM_011536635.2
XP_011534937.1
21
5688
XM_011536638.2
XP_011534940.1
17
4344
XM_017021159.1
XP_016876648.1
24
5778
XM_017021160.1
XP_016876649.1
22
5700
XM_017021161.1
XP_016876650.1
23
5653
XM_017021162.1
XP_016876651.1
25
5835
XM_017021163.1
XP_016876652.1
20
5502
XM_017021164.1
XP_016876653.1
20
5509
XM_017021165.1
XP_016876654.1
22
5578
XM_017021166.1
XP_016876655.1
23
5711
XM_017021167.1
XP_016876656.1
22
5586
XM_017021168.1
XP_016876657.1
22
5560
XM_017021169.1
XP_016876658.1
21
5713
XM_017021170.1
XP_016876659.1
21
6071
XM_017021171.1
XP_016876660.1
22
5644
XM_017021172.1
XP_016876661.1
21
5519
XM_017021173.1
XP_016876662.1
20
5646
XM_017021174.1
XP_016876663.1
20
5886
XM_017021175.1
XP_016876664.1
20
6004
XM_017021176.1
XP_016876665.1
25
5831
XM_017021177.1
XP_016876666.1
25
5820
XM_017021178.1
XP_016876667.1
23
5658
XM_017021179.1
XP_016876668.1
22
5561
XM_017021180.1
XP_016876669.1
26
5873
XM_017021181.1
XP_016876670.1
24
5778
XM_017021182.1
XP_016876671.1
23
5711
XM_017021183.1
XP_016876672.1
21
5519
XM_017021184.1
XP_016876673.1
25
5831
XM_017021185.1
XP_016876674.1
18
5859
XM_017021186.1
XP_016876675.1
23
5715
XM_017021187.1
XP_016876676.1
21
5581
XM_017021188.1
XP_016876677.1
20
5456
XM_017021189.1
XP_016876678.1
22
5565
XM_017021190.1
XP_016876679.1
23
5715
XM_017021191.1
XP_016876680.1
21
5523
XM_017021192.1
XP_016876681.1
20
5456
XM_017021193.1
XP_016876682.1
20
5454
XM_017021194.1
XP_016876683.1
23
5631
XM_017021195.1
XP_016876684.1
22
5568
XM_017021196.1
XP_016876685.1
22
5568
XM_017021197.1
XP_016876686.1
23
3978
XM_017021198.1
XP_016876687.1
20
3719
XM_017021199.1
XP_016876688.1
24
4020
XM_017021200.1
XP_016876689.1
20
3723
GenBank mRNA
AB007900.2
-
17
4994
AF090989.1
-
17
5194
AF090990.1
-
18
5257
AK122930.1
-
17
5796
AK302650.1
-
16
3446
BC146777.1
-
17
4994
9007226
CR936651.1
-
3
1003
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
actin cytoskeleton reorganization
regulation of GTPase activity
ephrin receptor signaling pathway
regulation of synaptic plasticity
regulation of axonogenesis
regulation of small GTPase mediated signal transduction
regulation of dendritic spine morphogenesis
activation of GTPase activity
molecular_function
GTPase activator activity
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0