Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERC2 SIRNA Assays ›
Gene Symbol:
ERC2
Gene Name:
ELKS/RAB6-interacting/CAST family member 2
Gene Aliases:
CAST, CAST1, ELKSL, SPBC110, Spc110
Chromosome Location:
Chr.3: 55508311 - 56468473 on Build GRCh38
UniGene ID:
Hs.476389
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s25036
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015576.2
NP_056391.1
2
446
NR_132749.1
2
446
XM_017006138.1
XP_016861627.1
2
460
XM_017006139.1
XP_016861628.1
2
460
XM_017006140.1
XP_016861629.1
2
459
XM_017006141.1
XP_016861630.1
2
474
XM_017006142.1
XP_016861631.1
2
474
XM_017006143.1
XP_016861632.1
2
459
XM_017006144.1
XP_016861633.1
2
459
XM_017006145.1
XP_016861634.1
2
459
XM_017006146.1
XP_016861635.1
2
474
XM_017006147.1
XP_016861636.1
2
459
XM_017006148.1
XP_016861637.1
2
474
XM_017006149.1
XP_016861638.1
2
533
XM_017006150.1
XP_016861639.1
2
474
XM_017006151.1
XP_016861640.1
2
474
XM_017006152.1
XP_016861641.1
2
459
XM_017006153.1
XP_016861642.1
2
459
XM_017006154.1
XP_016861643.1
2
459
XM_017006155.1
XP_016861644.1
2
556
XM_017006156.1
XP_016861645.1
2
474
XM_017006157.1
XP_016861646.1
2
459
XM_017006158.1
XP_016861647.1
2
474
XM_017006159.1
XP_016861648.1
2
513
GenBank mRNA
AB002376.2
-
2
446
BC111550.1
-
2
420
40008953
BC112391.1
-
2
420
40008962
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0