Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAI14 SIRNA Assays ›
Gene Symbol:
RAI14
Gene Name:
retinoic acid induced 14
Gene Aliases:
NORPEG, RAI13
Chromosome Location:
Chr.5: 34656328 - 34832612 on Build GRCh38
UniGene ID:
Hs.431400
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s25047
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RAI14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145520.1
NP_001138992.1
13, 14
1259
NM_001145521.1
NP_001138993.1
14, 15
1372
NM_001145522.1
NP_001138994.1
12, 13
1114
NM_001145523.1
NP_001138995.1
12, 13
1133
NM_001145525.1
NP_001138997.1
15, 16
1553
NM_015577.2
NP_056392.2
13, 14
1201
XM_006714469.2
XP_006714532.1
13, 14
1168
XM_011514016.2
XP_011512318.2
12, 13
1115
XM_011514017.2
XP_011512319.1
13, 14
1292
XM_011514018.1
XP_011512320.1
13, 14
1293
XM_011514019.1
XP_011512321.1
14, 15
1267
XM_011514020.1
XP_011512322.1
13, 14
1226
XM_011514021.1
XP_011512323.1
13, 14
1130
XM_011514022.1
XP_011512324.1
13, 14
1109
XM_011514023.1
XP_011512325.1
13, 14
1151
XM_011514024.1
XP_011512326.1
13, 14
1144
XM_011514025.2
XP_011512327.2
11, 12
1025
XM_017009334.1
XP_016864823.1
12, 13
1310
XM_017009335.1
XP_016864824.1
12, 13
1193
XM_017009336.1
XP_016864825.1
12, 13
1057
GenBank mRNA
AB037755.1
-
13, 14
1259
AF155135.1
-
13, 14
1116
AK297964.1
-
6, 7
551
AK308306.1
-
14, 15
1295
AK308455.1
-
4, 5
228
AK314379.1
-
12, 13
1003
AY317139.1
-
15, 16
1506
AY354204.2
-
12, 13
1133
BC052988.1
-
13, 14
1117
6164087
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0