Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPK1 SIRNA Assays ›
Gene Symbol:
TPK1
Gene Name:
thiamin pyrophosphokinase 1
Gene Aliases:
HTPK1, PP20, THMD5
Chromosome Location:
Chr.7: 144451941 - 144836424 on Build GRCh38
UniGene ID:
Hs.660232
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s25693
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042482.1
NP_001035947.1
5
319
NM_022445.3
NP_071890.2
5
319
XM_005249970.1
XP_005250027.1
5
297
XM_011516031.1
XP_011514333.1
5
320
XM_011516032.2
XP_011514334.1
5
456
XM_011516033.2
XP_011514335.1
5
326
XM_011516034.2
XP_011514336.1
5
383
XM_011516035.2
XP_011514337.1
5
320
XM_011516037.2
XP_011514339.1
4
567
XM_011516039.2
XP_011514341.1
4
502
XM_011516040.2
XP_011514342.1
5
320
XM_011516041.2
XP_011514343.1
4
548
XM_011516043.1
XP_011514345.1
5
320
XM_011516044.1
XP_011514346.1
3
222
XM_011516046.1
XP_011514348.1
5
320
XM_011516047.2
XP_011514349.1
2
1069
XM_011516048.1
XP_011514350.1
1
35
XM_011516050.1
XP_011514352.1
2
50
XM_017011969.1
XP_016867458.1
5
320
XM_017011970.1
XP_016867459.1
4
294
XM_017011971.1
XP_016867460.1
4
244
XM_017011972.1
XP_016867461.1
5
320
XM_017011973.1
XP_016867462.1
5
464
XM_017011974.1
XP_016867463.1
4
231
XM_017011975.1
XP_016867464.1
4
182
XM_017011976.1
XP_016867465.1
5
319
XM_017011977.1
XP_016867466.1
6
412
XM_017011978.1
XP_016867467.1
5
371
XM_017011979.1
XP_016867468.1
5
451
XM_017011982.1
XP_016867471.1
3
218
XM_017011983.1
XP_016867472.1
3
162
XM_017011984.1
XP_016867473.1
3
221
XM_017011986.1
XP_016867475.1
2
105
GenBank mRNA
AB028138.1
-
4
234
AF297710.1
-
5
306
AK026374.1
-
3
126
AK091736.1
-
5
258
AK128661.1
-
4
272
AK289652.1
-
5
294
AK308693.1
-
5
319
AY206415.1
-
5
301
BC040555.1
-
6
467
5269277
BC068460.1
-
5
298
5288626
BX446289.2
-
5
300
CR749248.1
-
3
306
DA135998.1
-
5
319
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
thiamine metabolic process
thiamine diphosphate biosynthetic process
phosphorylation
thiamine-containing compound metabolic process
thiamine diphosphokinase activity
ATP binding
kinase activity
thiamine binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0