Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP2C1 SIRNA Assays ›
          Gene Symbol:
          ATP2C1
          Gene Name:
          ATPase secretory pathway Ca2+ transporting 1
          Gene Aliases:
          ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
          Chromosome Location:
          Chr.3: 130850500 - 131016712 on Build GRCh38
          UniGene ID:
          Hs.584884
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s25717
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 562.00
          Your Price
          Online offer: 533.65
          533.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATP2C1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001485.2 NP_001001485.1 22, 23 2563
          NM_001001486.1 NP_001001486.1 21, 22 2240
          NM_001001487.1 NP_001001487.1 21, 22 2240
          NM_001199179.1 NP_001186108.1 22, 23 2469
          NM_001199180.1 NP_001186109.1 21, 22 2458
          NM_001199181.1 NP_001186110.1 21, 22 2458
          NM_001199182.1 NP_001186111.1 20, 21 2341
          NM_001199183.1 NP_001186112.1 21, 22 2331
          NM_001199184.1 NP_001186113.1 21, 22 2331
          NM_001199185.1 NP_001186114.1 21, 22 2382
          NM_014382.3 NP_055197.2 21, 22 2382
          XM_005247354.2 XP_005247411.1 21, 22 2483
          XM_005247355.2 XP_005247412.1 22, 23 2610
          XM_005247356.2 XP_005247413.1 22, 23 2363
          XM_005247358.2 XP_005247415.1 21, 22 2187
          XM_011512686.2 XP_011510988.1 12, 13 1292
          XM_017006164.1 XP_016861653.1 22, 23 2271
          GenBank mRNA AB037768.1 - 22, 23 2470
          AB209265.1 - 10, 11 2230
          AF181120.1 - 21, 22 2295
          AF181121.1 - 21, 22 2295
          AF189723.2 - 21, 22 2214
          AF225981.1 - 21, 22 2214
          AJ010953.1 - 7, 8 792
          AK001684.1 - 9, 10 990
          AK074692.1 - 19, 20 1921
          AK296470.1 - 21, 22 2385
          AK299945.1 - 20, 21 2097
          AK304175.1 - 21, 22 2187
          AK304374.1 - 21, 22 2214
          AK314342.1 - 21, 22 2244
          AY268374.1 - 21, 22 2189
          AY268375.1 - 21, 22 2189
          BC028139.1 - 21, 22 2252 5215224

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00205122_m1

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          cellular calcium ion homeostasis
          signal transduction
          epidermis development
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          actin cytoskeleton reorganization
          Golgi calcium ion homeostasis
          Golgi calcium ion transport
          ion transmembrane transport
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          calcium ion transmembrane transport
          signal transducer activity
          calcium-transporting ATPase activity
          ATP binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0