Search
Search

B3GAT1| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018644.3 | NP_061114.2 | 7 | 1857 | ||
| NM_054025.2 | NP_473366.1 | 6 | 1989 | |||
| XM_005271506.3 | XP_005271563.1 | 7 | 1908 | |||
| XM_011542751.2 | XP_011541053.1 | 8 | 2120 | |||
| XM_017017551.1 | XP_016873040.1 | 7 | 3682 | |||
| XM_017017552.1 | XP_016873041.1 | 5 | 2444 | |||
| GenBank mRNA | AL834455.1 | - | 4 | 1165 | ||
| BC010466.1 | - | 6 | 1989 | 4157589 | ||