Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DROSHA SIRNA Assays ›
          Gene Symbol:
          DROSHA
          Gene Name:
          drosha ribonuclease III
          Gene Aliases:
          ETOHI2, HSA242976, RANSE3L, RN3, RNASE3L, RNASEN
          Chromosome Location:
          Chr.5: 31400494 - 31532175 on Build GRCh38
          UniGene ID:
          Hs.97997
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s26492
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DROSHA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100412.1 NP_001093882.1 29, 30 3849
          NM_013235.4 NP_037367.3 29, 30 3884
          XM_005248291.3 XP_005248348.1 30, 31 3960
          XM_005248292.3 XP_005248349.1 30, 31 3936
          XM_005248293.3 XP_005248350.1 29, 30 3867
          XM_005248294.3 XP_005248351.1 28, 29 3756
          XM_011514033.2 XP_011512335.1 30, 31 3960
          XM_017009399.1 XP_016864888.1 29, 30 3843
          XM_017009400.1 XP_016864889.1 29, 30 3825
          XM_017009401.1 XP_016864890.1 28, 29 3732
          XM_017009402.1 XP_016864891.1 18, 19 1975
          GenBank mRNA AF116910.1 - 14, 15 1413
          AF189011.1 - 29, 30 3770
          AK001121.1 - 18, 19 1719
          AK026563.1 - 9, 10 771
          BC024261.1 - 17, 18 1645
          BC041162.1 - 16, 17 1570
          BC054003.1 - 28, 29 3661 6148714
          BX647724.1 - 29, 30 3752

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00294820_m1

          Panther Classification:

          Molecular Function -

          endoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          miRNA metabolic process
          positive regulation of gene expression
          rRNA catabolic process
          production of siRNA involved in RNA interference
          gene silencing by RNA
          primary miRNA processing
          pre-miRNA processing
          ribosome biogenesis
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          RNA phosphodiester bond hydrolysis, endonucleolytic
          regulation of miRNA metabolic process
          lipopolysaccharide binding
          double-stranded RNA binding
          ribonuclease III activity
          protein binding
          protein homodimerization activity
          poly(A) RNA binding
          metal ion binding
          primary miRNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline