Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASPH SIRNA Assays ›
Gene Symbol:
ASPH
Gene Name:
aspartate beta-hydroxylase
Gene Aliases:
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location:
Chr.8: 61500556 - 61714640 on Build GRCh38
UniGene ID:
Hs.332422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s270
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASPH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164750.1
NP_001158222.1
2
304
NM_001164751.1
NP_001158223.1
2
304
NM_001164752.1
NP_001158224.1
2
304
NM_001164753.1
NP_001158225.1
2
304
NM_001164754.1
NP_001158226.1
2
499
NM_001164755.1
NP_001158227.1
2
499
NM_001164756.1
NP_001158228.1
2
499
NM_004318.3
NP_004309.2
2
499
NM_020164.4
NP_064549.1
2
304
NM_032466.3
NP_115855.1
2
499
NM_032467.3
NP_115856.1
2
304
NM_032468.4
NP_115857.1
2
304
XM_005251235.2
XP_005251292.1
2
468
XM_005251236.2
XP_005251293.1
2
468
XM_005251238.2
XP_005251295.1
2
468
XM_005251239.2
XP_005251296.1
2
468
XM_005251240.1
XP_005251297.1
2
246
XM_005251242.2
XP_005251299.1
2
468
XM_005251243.2
XP_005251300.1
2
468
XM_005251244.1
XP_005251301.1
2
246
XM_005251246.2
XP_005251303.1
2
468
XM_005251247.2
XP_005251304.1
2
468
XM_005251248.1
XP_005251305.1
2
154
XM_005251250.2
XP_005251307.1
2
468
XM_017013419.1
XP_016868908.1
2
468
XM_017013420.1
XP_016868909.1
2
468
XM_017013421.1
XP_016868910.1
2
468
XM_017013422.1
XP_016868911.1
2
468
XM_017013423.1
XP_016868912.1
2
468
XM_017013424.1
XP_016868913.1
2
468
XM_017013425.1
XP_016868914.1
2
468
XM_017013426.1
XP_016868915.1
2
246
XM_017013427.1
XP_016868916.1
2
246
XM_017013428.1
XP_016868917.1
2
468
XM_017013429.1
XP_016868918.1
2
468
XM_017013430.1
XP_016868919.1
2
468
XM_017013431.1
XP_016868920.1
2
154
XM_017013432.1
XP_016868921.1
2
468
XM_017013433.1
XP_016868922.1
2
154
XM_017013434.1
XP_016868923.1
2
468
XM_017013435.1
XP_016868924.1
2
246
XM_017013436.1
XP_016868925.1
2
468
XM_017013437.1
XP_016868926.1
2
468
XM_017013438.1
XP_016868927.1
2
468
XM_017013439.1
XP_016868928.1
2
154
XM_017013440.1
XP_016868929.1
2
246
XM_017013441.1
XP_016868930.1
2
468
XM_017013442.1
XP_016868931.1
2
468
XM_017013443.1
XP_016868932.1
2
468
XM_017013444.1
XP_016868933.1
2
154
XM_017013445.1
XP_016868934.1
2
154
XM_017013446.1
XP_016868935.1
2
154
XM_017013447.1
XP_016868936.1
2
154
GenBank mRNA
AF184241.1
-
2
184
AF224468.1
-
2
254
AF224469.1
-
2
229
AF289489.1
-
2
241
AF306765.1
-
2
225
AK291836.1
-
2
312
AK295528.1
-
2
268
AK298577.1
-
3
420
AK304314.1
-
2
260
BC015518.2
-
2
418
BC025236.1
-
2
427
BC066929.1
-
2
395
BC142967.1
-
2
376
BC144362.1
-
2
234
BC144363.1
-
2
235
CR996516.1
-
2
499
DA136318.1
-
2
468
DN993915.1
-
2
312
EU718494.1
-
2
307
FJ461473.1
-
2
230
S83325.1
-
2
241
U03109.1
-
2
307
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00188872_m1
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
pattern specification process
negative regulation of cell proliferation
regulation of protein stability
ion transmembrane transport
limb morphogenesis
peptidyl-aspartic acid hydroxylation
positive regulation of proteolysis
oxidation-reduction process
palate development
face morphogenesis
activation of cysteine-type endopeptidase activity
regulation of protein depolymerization
regulation of cardiac conduction
peptide-aspartate beta-dioxygenase activity
structural molecule activity
calcium ion binding
protein binding
structural constituent of muscle
electron carrier activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0