Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD74 SIRNA Assays ›
          Gene Symbol:
          CD74
          Gene Name:
          CD74 molecule
          Gene Aliases:
          DHLAG, HLADG, II, Ia-GAMMA
          Chromosome Location:
          Chr.5: 150401637 - 150412936 on Build GRCh38
          UniGene ID:
          Hs.436568
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2715

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD74
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025158.2 NP_001020329.1 1 254
          NM_001025159.2 NP_001020330.1 1 254
          NM_004355.3 NP_004346.1 1 254
          XM_017010089.1 XP_016865578.1 1 230
          XM_017010090.1 XP_016865579.1 1 230
          GenBank mRNA AK292076.1 - 1 69
          AK297889.1 - 1 69
          AK300669.1 - 1 69
          AK308929.1 - 1 69
          BC018726.1 - 1 78 4853578
          BC024272.1 - 1 87 4763768
          BF796148.1 - 1 57
          BT019505.1 - 1 67
          DC354760.1 - 1 254
          K01144.1 - 1 118
          X00497.1 - 1 74

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          prostaglandin biosynthetic process
          positive regulation of cytokine-mediated signaling pathway
          positive regulation of dendritic cell antigen processing and presentation
          negative regulation of peptide secretion
          positive regulation of type 2 immune response
          negative regulation of mature B cell apoptotic process
          protein complex assembly
          intracellular protein transport
          defense response
          signal transduction
          cell proliferation
          immunoglobulin mediated immune response
          antigen processing and presentation of endogenous antigen
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          positive regulation of B cell proliferation
          macrophage migration inhibitory factor signaling pathway
          regulation of macrophage activation
          negative regulation of apoptotic process
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          T cell selection
          positive thymic T cell selection
          negative thymic T cell selection
          negative regulation of T cell differentiation
          positive regulation of T cell differentiation
          positive regulation of fibroblast proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte migration
          chaperone mediated protein folding requiring cofactor
          positive regulation of macrophage cytokine production
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of neutrophil chemotaxis
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          beta-amyloid binding
          cytokine receptor activity
          protein binding
          cytokine binding
          MHC class II protein complex binding
          macrophage migration inhibitory factor binding
          MHC class II protein binding
          MHC class II protein binding, via antigen binding groove
          identical protein binding
          protein binding involved in protein folding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-sg7j2:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0