Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MECR SIRNA Assays ›
          Gene Symbol:
          MECR
          Gene Name:
          mitochondrial trans-2-enoyl-CoA reductase
          Gene Aliases:
          CGI-63, FASN2B, NRBF1
          Chromosome Location:
          Chr.1: 29192657 - 29230958 on Build GRCh38
          UniGene ID:
          Hs.183646
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s27434

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MECR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024732.2 NP_001019903.2 5, 6 837
          NM_016011.3 NP_057095.3 5, 6 710
          XM_005245885.2 XP_005245942.1 6, 7 760
          XM_005245887.2 XP_005245944.1 7, 8 1006
          XM_011541539.1 XP_011539841.1 6, 7 760
          XM_011541540.1 XP_011539842.1 5, 6 699
          XM_011541541.1 XP_011539843.1 6, 7 760
          XM_011541543.1 XP_011539845.1 6, 7 869
          XM_011541545.1 XP_011539847.1 7, 8 1033
          XM_011541546.1 XP_011539848.1 5, 6 822
          XM_011541547.1 XP_011539849.1 8, 9 1155
          XM_011541549.1 XP_011539851.1 7, 8 1050
          XM_011541550.1 XP_011539852.1 7, 8 905
          XM_011541553.1 XP_011539855.1 7, 8 906
          XM_011541554.1 XP_011539856.1 6, 7 670
          XM_017001411.1 XP_016856900.1 7, 8 1112
          XM_017001412.1 XP_016856901.1 7, 8 1027
          XM_017001413.1 XP_016856902.1 7, 8 781
          XM_017001414.1 XP_016856903.1 8, 9 1028
          XM_017001415.1 XP_016856904.1 7, 8 899
          XM_017001416.1 XP_016856905.1 7, 8 906
          GenBank mRNA AF151821.1 - 5, 6 684
          AK025830.1 - 5, 6 676
          AK095099.1 - 5, 6 803
          AK226084.1 - 5, 6 784
          AK300889.1 - 5, 6 674
          AK315274.1 - 5, 6 697
          BC001419.2 - 5, 6 681 3139846
          CR456703.1 - 5, 6 658

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid metabolic process
          fatty acid biosynthetic process
          oxidation-reduction process
          zinc ion binding
          trans-2-enoyl-CoA reductase (NADPH) activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0