Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LEF1 SIRNA Assays ›
          Gene Symbol:
          LEF1
          Gene Name:
          lymphoid enhancer binding factor 1
          Gene Aliases:
          LEF-1, TCF10, TCF1ALPHA, TCF7L3
          Chromosome Location:
          Chr.4: 108047545 - 108168956 on Build GRCh38
          UniGene ID:
          Hs.743478
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s27617

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LEF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130713.2 NP_001124185.1 8 2210
          NM_001130714.2 NP_001124186.1 8 2210
          NM_001166119.1 NP_001159591.1 8 1334
          NM_016269.4 NP_057353.1 9 2294
          XM_005263046.3 XP_005263103.1 9 1298
          XM_005263047.1 XP_005263104.1 9 922
          XM_005263048.1 XP_005263105.1 8 838
          XM_006714233.1 XP_006714296.1 9 999
          GenBank mRNA AF086339.1 - 3 359
          AF198532.1 - 9 1105
          AF288571.1 - 9 1759
          AF294627.1 - 9 1185
          AK225772.1 - 8 2098
          AK294395.1 - 8 1052
          AK303143.1 - 7 900
          AK303272.1 - 8 1258
          AL049409.1 - 2 117
          BC040559.1 - 8 1043 5269696
          BC050632.2 - 9 1196 6054813

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          patterning of blood vessels
          osteoblast differentiation
          neural crest cell migration
          somitogenesis
          kidney development
          epithelial to mesenchymal transition
          sprouting angiogenesis
          transcription from RNA polymerase II promoter
          Wnt signaling pathway, calcium modulating pathway
          positive regulation of cell proliferation
          response to lithium ion
          positive regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          Wnt signaling pathway
          regulation of striated muscle tissue development
          dentate gyrus development
          hypothalamus development
          forebrain radial glial cell differentiation
          forebrain neuroblast division
          forebrain neuron differentiation
          formation of radial glial scaffolds
          regulation of cell-cell adhesion
          neutrophil differentiation
          positive regulation of cell growth
          embryonic limb morphogenesis
          positive regulation of cell migration
          BMP signaling pathway
          positive regulation of granulocyte differentiation
          mammary gland development
          organ regeneration
          negative regulation of interleukin-13 production
          negative regulation of interleukin-4 production
          negative regulation of interleukin-5 production
          T cell receptor V(D)J recombination
          B cell proliferation
          odontogenesis of dentin-containing tooth
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of DNA binding
          steroid hormone mediated signaling pathway
          tongue development
          skin development
          positive regulation by host of viral transcription
          histone H3 acetylation
          histone H4 acetylation
          T-helper 1 cell differentiation
          negative regulation of striated muscle tissue development
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          alpha-beta T cell differentiation
          eye pigmentation
          paraxial mesoderm formation
          muscle fiber development
          sensory perception of taste
          palate development
          anatomical structure regression
          canonical Wnt signaling pathway
          face morphogenesis
          cell chemotaxis
          apoptotic process involved in morphogenesis
          chorio-allantoic fusion
          trachea gland development
          cellular response to cytokine stimulus
          cellular response to interleukin-4
          positive regulation of cell proliferation in bone marrow
          negative regulation of apoptotic process in bone marrow
          odontoblast differentiation
          negative regulation of estrogen receptor binding
          negative regulation of canonical Wnt signaling pathway
          apoptotic process involved in patterning of blood vessels
          beta-catenin-TCF complex assembly
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          beta-catenin binding
          transcription factor binding
          DNA binding, bending
          estrogen receptor activity
          estrogen receptor binding
          enhancer binding
          histone binding
          histone deacetylase binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          gamma-catenin binding
          armadillo repeat domain binding
          C2H2 zinc finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0